Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57617968-57618241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCOM1
Entrez Gene ID
145781
Gene Name
GRINL1A complex locus 1
Gene Aliases
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Gene Symbol
MYZAP
Entrez Gene ID
100820829
Gene Name
myocardial zonula adherens protein
Gene Aliases
GCOM1, Gup, MYOZAP
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57685364 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3502692 c.271G>A p.G91R Chr.15: 57618141-57618141 on Build GRCh38
6142626 c.193A>G p.R65G Chr.15: 57618063-57618063 on Build GRCh38
6001835 c.229C>T p.R77* Chr.15: 57618099-57618099 on Build GRCh38
3502689 c.241C>T p.Q81* Chr.15: 57618111-57618111 on Build GRCh38
3502686 c.230G>A p.R77Q Chr.15: 57618100-57618100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779445376 Chr.15: 57618115-57618115 on Build GRCh38
rs767599641 Chr.15: 57618001-57618001 on Build GRCh38
rs750678507 Chr.15: 57618140-57618140 on Build GRCh38
rs992189342 Chr.15: 57618106-57618106 on Build GRCh38
rs765756258 Chr.15: 57618189-57618189 on Build GRCh38
rs754163797 Chr.15: 57618013-57618013 on Build GRCh38
rs377330516 Chr.15: 57618063-57618063 on Build GRCh38
rs762406940 Chr.15: 57618185-57618185 on Build GRCh38
rs762687142 Chr.15: 57618204-57618204 on Build GRCh38
rs775462379 Chr.15: 57618160-57618160 on Build GRCh38
rs766019015 Chr.15: 57618201-57618201 on Build GRCh38
rs761090068 Chr.15: 57618083-57618083 on Build GRCh38
rs755984426 Chr.15: 57618214-57618214 on Build GRCh38
rs751017394 Chr.15: 57618119-57618119 on Build GRCh38
rs374058925 Chr.15: 57618053-57618053 on Build GRCh38
rs147173331 Chr.15: 57618043-57618043 on Build GRCh38
rs767006038 Chr.15: 57618211-57618211 on Build GRCh38
rs141271435 Chr.15: 57618079-57618079 on Build GRCh38
rs957471267 Chr.15: 57618005-57618005 on Build GRCh38
rs866578359 Chr.15: 57618100-57618100 on Build GRCh38
rs375935328 Chr.15: 57618216-57618216 on Build GRCh38
rs769550734 Chr.15: 57618069-57618069 on Build GRCh38
rs561682165 Chr.15: 57618208-57618208 on Build GRCh38
rs572750557 Chr.15: 57618156-57618156 on Build GRCh38
rs758057825 Chr.15: 57618114-57618114 on Build GRCh38
rs370920807 Chr.15: 57618064-57618064 on Build GRCh38
rs754906541 Chr.15: 57618123-57618123 on Build GRCh38
rs777318558 Chr.15: 57618179-57618179 on Build GRCh38
rs199944789 Chr.15: 57618026-57618026 on Build GRCh38
rs781138834 Chr.15: 57618125-57618125 on Build GRCh38
rs909467384 Chr.15: 57618112-57618112 on Build GRCh38
rs750168470 Chr.15: 57618099-57618099 on Build GRCh38
rs754277372 Chr.15: 57618093-57618093 on Build GRCh38
rs773655794 Chr.15: 57618194-57618194 on Build GRCh38
rs746101335 Chr.15: 57618149-57618149 on Build GRCh38
rs919404550 Chr.15: 57618169-57618169 on Build GRCh38
rs878883998 Chr.15: 57618003-57618005 on Build GRCh38
rs760759277 Chr.15: 57618161-57618161 on Build GRCh38
rs867428039 Chr.15: 57618148-57618148 on Build GRCh38
rs768583581 Chr.15: 57618059-57618059 on Build GRCh38
rs752686523 Chr.15: 57618212-57618212 on Build GRCh38
rs200977499 Chr.15: 57618011-57618011 on Build GRCh38
rs557151193 Chr.15: 57618041-57618041 on Build GRCh38
rs539562560 Chr.15: 57618075-57618075 on Build GRCh38
rs762317142 Chr.15: 57618095-57618095 on Build GRCh38
rs761228518 Chr.15: 57618003-57618003 on Build GRCh38
rs550847444 Chr.15: 57617995-57617995 on Build GRCh38
rs942240097 Chr.15: 57618129-57618129 on Build GRCh38
rs778660333 Chr.15: 57618039-57618039 on Build GRCh38
rs768596133 Chr.15: 57618171-57618171 on Build GRCh38
rs765666243 Chr.15: 57618096-57618096 on Build GRCh38
rs777527520 Chr.15: 57618147-57618147 on Build GRCh38
rs779614211 Chr.15: 57618050-57618050 on Build GRCh38
rs200548712 Chr.15: 57618196-57618196 on Build GRCh38
rs754552037 Chr.15: 57618210-57618210 on Build GRCh38
rs764620781 Chr.15: 57618088-57618088 on Build GRCh38
rs147715722 Chr.15: 57618068-57618068 on Build GRCh38
rs753208716 Chr.15: 57618002-57618002 on Build GRCh38
rs138712430 Chr.15: 57618135-57618135 on Build GRCh38

View Full Product Details