Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5772431-5772656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMP1
Entrez Gene ID
79956
Gene Name
endoplasmic reticulum metallopeptidase 1
Gene Aliases
FXNA, KIAA1815, bA207C16.3
UniGene
Hs.591078
Gene Chromosome Location
Chr.9: 5764061-5867091 on Build GRCh38
Transcripts
NM_001206557.1 NM_020829.3 XM_005251523.3 XM_011517967.2 XM_011517968.2 XM_017014934.1 XM_017014935.1 XM_017014936.1

Gene Symbol
RIC1
Entrez Gene ID
57589
Gene Name
RIC1 homolog, RAB6A GEF complex partner 1
Gene Aliases
CIP150, KIAA1432, bA207C16.1
UniGene
Hs.211520
Gene Chromosome Location
Chr.9: 5629030-5778633 on Build GRCh38
Transcripts
NM_001206557.1 NM_020829.3 XM_005251523.3 XM_011517967.2 XM_011517968.2 XM_017014934.1 XM_017014935.1 XM_017014936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7000669 c.3404C>A p.A1135D Chr.9: 5772588-5772588 on Build GRCh38
3657681 c.3429C>T p.S1143S Chr.9: 5772613-5772613 on Build GRCh38
7274499 c.3424A>G p.S1142G Chr.9: 5772608-5772608 on Build GRCh38
5359608 c.3398G>C p.G1133A Chr.9: 5772582-5772582 on Build GRCh38
6275461 c.3446G>T p.W1149L Chr.9: 5772630-5772630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772020351 Chr.9: 5772614-5772614 on Build GRCh38
rs746867743 Chr.9: 5772609-5772609 on Build GRCh38
rs10124363 Chr.9: 5772550-5772550 on Build GRCh38
rs752627971 Chr.9: 5772591-5772591 on Build GRCh38
rs41280735 Chr.9: 5772537-5772537 on Build GRCh38
rs182033922 Chr.9: 5772528-5772528 on Build GRCh38
rs189532070 Chr.9: 5772484-5772484 on Build GRCh38
rs769891241 Chr.9: 5772616-5772616 on Build GRCh38
rs766106883 Chr.9: 5772556-5772556 on Build GRCh38
rs778574444 Chr.9: 5772572-5772572 on Build GRCh38
rs368434389 Chr.9: 5772552-5772552 on Build GRCh38
rs758435306 Chr.9: 5772598-5772598 on Build GRCh38
rs111546732 Chr.9: 5772539-5772539 on Build GRCh38
rs368306879 Chr.9: 5772535-5772535 on Build GRCh38
rs374377906 Chr.9: 5772529-5772529 on Build GRCh38
rs1010649281 Chr.9: 5772478-5772478 on Build GRCh38
rs932377692 Chr.9: 5772490-5772490 on Build GRCh38
rs1049981012 Chr.9: 5772505-5772505 on Build GRCh38
rs372066029 Chr.9: 5772519-5772519 on Build GRCh38
rs771930773 Chr.9: 5772517-5772517 on Build GRCh38
rs138589800 Chr.9: 5772625-5772625 on Build GRCh38
rs753274442 Chr.9: 5772560-5772560 on Build GRCh38
rs769143055 Chr.9: 5772536-5772536 on Build GRCh38
rs754602568 Chr.9: 5772565-5772565 on Build GRCh38
rs922857267 Chr.9: 5772619-5772619 on Build GRCh38
rs746031028 Chr.9: 5772526-5772526 on Build GRCh38
rs904859743 Chr.9: 5772488-5772488 on Build GRCh38
rs10975276 Chr.9: 5772465-5772465 on Build GRCh38
rs777641108 Chr.9: 5772523-5772523 on Build GRCh38
rs976551648 Chr.9: 5772474-5772474 on Build GRCh38
rs777394864 Chr.9: 5772608-5772608 on Build GRCh38
rs747952981 Chr.9: 5772516-5772516 on Build GRCh38
rs984152474 Chr.9: 5772543-5772543 on Build GRCh38

View Full Product Details