Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 77196141-77196414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC146
Entrez Gene ID
57639
Gene Name
coiled-coil domain containing 146
Gene Aliases
-
UniGene
Hs.113940
Gene Chromosome Location
Chr.7: 77122617-77295204 on Build GRCh38
Transcripts
NM_020879.2 NM_006682.2

Gene Symbol
FGL2
Entrez Gene ID
10875
Gene Name
fibrinogen like 2
Gene Aliases
T49, pT49
UniGene
Hs.520989
Gene Chromosome Location
Chr.7: 77193371-77199833 on Build GRCh38
Transcripts
NM_020879.2 NM_006682.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3882483 c.1238T>C p.V413A Chr.7: 77196361-77196361 on Build GRCh38
3394892 c.1198G>C p.G400R Chr.7: 77196401-77196401 on Build GRCh38
3882482 c.1241G>T p.S414I Chr.7: 77196358-77196358 on Build GRCh38
6663869 c.1191delA p.K397fs*17 Chr.7: 77196408-77196408 on Build GRCh38
6663873 c.1205G>A p.R402H Chr.7: 77196394-77196394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200580249 Chr.7: 77196348-77196348 on Build GRCh38
rs750223519 Chr.7: 77196281-77196281 on Build GRCh38
rs1027070056 Chr.7: 77196217-77196217 on Build GRCh38
rs780134047 Chr.7: 77196297-77196297 on Build GRCh38
rs755727408 Chr.7: 77196223-77196223 on Build GRCh38
rs1016867186 Chr.7: 77196365-77196365 on Build GRCh38
rs139238249 Chr.7: 77196242-77196242 on Build GRCh38
rs771487735 Chr.7: 77196372-77196372 on Build GRCh38
rs769386251 Chr.7: 77196347-77196347 on Build GRCh38
rs904781594 Chr.7: 77196218-77196218 on Build GRCh38
rs746556008 Chr.7: 77196303-77196303 on Build GRCh38
rs954263611 Chr.7: 77196264-77196264 on Build GRCh38
rs373117921 Chr.7: 77196247-77196247 on Build GRCh38
rs575304336 Chr.7: 77196215-77196215 on Build GRCh38
rs747864954 Chr.7: 77196345-77196345 on Build GRCh38
rs187050930 Chr.7: 77196206-77196206 on Build GRCh38
rs182762397 Chr.7: 77196165-77196165 on Build GRCh38
rs1028446662 Chr.7: 77196260-77196260 on Build GRCh38
rs766846655 Chr.7: 77196239-77196239 on Build GRCh38
rs763305655 Chr.7: 77196366-77196366 on Build GRCh38
rs768221638 Chr.7: 77196309-77196309 on Build GRCh38
rs900063718 Chr.7: 77196202-77196202 on Build GRCh38
rs894211426 Chr.7: 77196187-77196187 on Build GRCh38
rs369065862 Chr.7: 77196338-77196338 on Build GRCh38
rs377447118 Chr.7: 77196324-77196324 on Build GRCh38
rs758447764 Chr.7: 77196292-77196292 on Build GRCh38
rs201977884 Chr.7: 77196371-77196371 on Build GRCh38
rs753777784 Chr.7: 77196257-77196257 on Build GRCh38
rs368687438 Chr.7: 77196250-77196250 on Build GRCh38
rs756251312 Chr.7: 77196251-77196251 on Build GRCh38
rs377459664 Chr.7: 77196241-77196241 on Build GRCh38
rs3093279 Chr.7: 77196166-77196166 on Build GRCh38

View Full Product Details