Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2799021-2799294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP4
Entrez Gene ID
2288
Gene Name
FK506 binding protein 4
Gene Aliases
FKBP51, FKBP52, FKBP59, HBI, Hsp56, PPIase, p52
UniGene
Hs.524183
Gene Chromosome Location
Chr.12: 2794942-2805423 on Build GRCh38
Transcripts
NM_002014.3 XM_011520929.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7368327 c.591C>A p.N197K Chr.12: 2799164-2799164 on Build GRCh38
6664264 c.543G>A p.K181K Chr.12: 2799116-2799116 on Build GRCh38
7368326 c.571G>C p.E191Q Chr.12: 2799144-2799144 on Build GRCh38
5798718 c.558G>A p.R186R Chr.12: 2799131-2799131 on Build GRCh38
6664262 c.655G>A p.V219M Chr.12: 2799228-2799228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780449875 Chr.12: 2799242-2799242 on Build GRCh38
rs1011831601 Chr.12: 2799101-2799101 on Build GRCh38
rs374042199 Chr.12: 2799114-2799114 on Build GRCh38
rs1031996000 Chr.12: 2799090-2799090 on Build GRCh38
rs768397372 Chr.12: 2799072-2799075 on Build GRCh38
rs750117425 Chr.12: 2799089-2799089 on Build GRCh38
rs776764262 Chr.12: 2799179-2799179 on Build GRCh38
rs760628848 Chr.12: 2799104-2799104 on Build GRCh38
rs761886074 Chr.12: 2799190-2799190 on Build GRCh38
rs369225968 Chr.12: 2799043-2799043 on Build GRCh38
rs747817842 Chr.12: 2799072-2799072 on Build GRCh38
rs769891493 Chr.12: 2799268-2799268 on Build GRCh38
rs747525966 Chr.12: 2799160-2799160 on Build GRCh38
rs776622503 Chr.12: 2799107-2799107 on Build GRCh38
rs776164317 Chr.12: 2799097-2799097 on Build GRCh38
rs746239939 Chr.12: 2799085-2799085 on Build GRCh38
rs902238548 Chr.12: 2799051-2799051 on Build GRCh38
rs780740336 Chr.12: 2799129-2799129 on Build GRCh38
rs369216531 Chr.12: 2799113-2799113 on Build GRCh38
rs748176412 Chr.12: 2799263-2799263 on Build GRCh38
rs751370254 Chr.12: 2799203-2799203 on Build GRCh38
rs147640045 Chr.12: 2799092-2799092 on Build GRCh38
rs1042734137 Chr.12: 2799216-2799216 on Build GRCh38
rs773444295 Chr.12: 2799200-2799200 on Build GRCh38
rs373445860 Chr.12: 2799068-2799068 on Build GRCh38
rs200255491 Chr.12: 2799225-2799225 on Build GRCh38
rs747396122 Chr.12: 2799243-2799243 on Build GRCh38
rs758199458 Chr.12: 2799116-2799116 on Build GRCh38
rs751486111 Chr.12: 2799128-2799128 on Build GRCh38
rs759514100 Chr.12: 2799206-2799206 on Build GRCh38
rs56196860 Chr.12: 2799164-2799164 on Build GRCh38
rs757016850 Chr.12: 2799239-2799239 on Build GRCh38
rs772597983 Chr.12: 2799153-2799153 on Build GRCh38
rs376456473 Chr.12: 2799148-2799148 on Build GRCh38
rs769944304 Chr.12: 2799199-2799199 on Build GRCh38
rs1017820517 Chr.12: 2799126-2799126 on Build GRCh38
rs55672055 Chr.12: 2799139-2799139 on Build GRCh38
rs774851464 Chr.12: 2799080-2799080 on Build GRCh38
rs202109074 Chr.12: 2799259-2799259 on Build GRCh38
rs147940690 Chr.12: 2799055-2799055 on Build GRCh38
rs762081103 Chr.12: 2799108-2799108 on Build GRCh38
rs141855441 Chr.12: 2799138-2799138 on Build GRCh38
rs781562456 Chr.12: 2799262-2799262 on Build GRCh38
rs777530787 Chr.12: 2799236-2799236 on Build GRCh38
rs927155815 Chr.12: 2799130-2799130 on Build GRCh38
rs909199883 Chr.12: 2799073-2799073 on Build GRCh38
rs747679046 Chr.12: 2799133-2799133 on Build GRCh38
rs780610678 Chr.12: 2799070-2799070 on Build GRCh38
rs371138035 Chr.12: 2799201-2799201 on Build GRCh38
rs144527066 Chr.12: 2799228-2799228 on Build GRCh38
rs558632124 Chr.12: 2799076-2799076 on Build GRCh38
rs376058719 Chr.12: 2799105-2799105 on Build GRCh38
rs369674741 Chr.12: 2799119-2799119 on Build GRCh38
rs143820830 Chr.12: 2799238-2799238 on Build GRCh38
rs755711294 Chr.12: 2799230-2799230 on Build GRCh38
rs746443863 Chr.12: 2799060-2799060 on Build GRCh38
rs766142249 Chr.12: 2799202-2799202 on Build GRCh38

View Full Product Details