Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108518500-108518743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FICD
Entrez Gene ID
11153
Gene Name
FIC domain containing
Gene Aliases
HIP13, HYPE, UNQ3041
UniGene
Hs.661891
Gene Chromosome Location
Chr.12: 108515274-108519603 on Build GRCh38
Transcripts
NM_007076.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7282178 c.560T>C p.V187A Chr.12: 108518658-108518658 on Build GRCh38
4166028 c.475G>A p.A159T Chr.12: 108518573-108518573 on Build GRCh38
161071 c.611A>C p.K204T Chr.12: 108518709-108518709 on Build GRCh38
3356279 c.551T>C p.L184P Chr.12: 108518649-108518649 on Build GRCh38
1605523 c.630G>T p.K210N Chr.12: 108518728-108518728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143888209 Chr.12: 108518578-108518578 on Build GRCh38
rs142079606 Chr.12: 108518657-108518657 on Build GRCh38
rs776804151 Chr.12: 108518630-108518630 on Build GRCh38
rs576372988 Chr.12: 108518603-108518603 on Build GRCh38
rs755373703 Chr.12: 108518706-108518706 on Build GRCh38
rs755964034 Chr.12: 108518671-108518671 on Build GRCh38
rs777122526 Chr.12: 108518548-108518548 on Build GRCh38
rs781738865 Chr.12: 108518708-108518708 on Build GRCh38
rs778111324 Chr.12: 108518638-108518638 on Build GRCh38
rs762582420 Chr.12: 108518569-108518569 on Build GRCh38
rs536188515 Chr.12: 108518651-108518651 on Build GRCh38
rs747512205 Chr.12: 108518617-108518617 on Build GRCh38
rs542666459 Chr.12: 108518522-108518522 on Build GRCh38
rs762741729 Chr.12: 108518641-108518641 on Build GRCh38
rs750615173 Chr.12: 108518693-108518693 on Build GRCh38
rs759530176 Chr.12: 108518647-108518647 on Build GRCh38
rs140133453 Chr.12: 108518539-108518539 on Build GRCh38
rs765955642 Chr.12: 108518642-108518642 on Build GRCh38
rs758453316 Chr.12: 108518602-108518602 on Build GRCh38
rs752117018 Chr.12: 108518686-108518688 on Build GRCh38
rs764956334 Chr.12: 108518554-108518554 on Build GRCh38
rs747107133 Chr.12: 108518704-108518704 on Build GRCh38
rs758309364 Chr.12: 108518547-108518547 on Build GRCh38
rs988080196 Chr.12: 108518557-108518557 on Build GRCh38
rs558604994 Chr.12: 108518634-108518634 on Build GRCh38
rs776003201 Chr.12: 108518544-108518544 on Build GRCh38
rs781087797 Chr.12: 108518524-108518524 on Build GRCh38
rs369383570 Chr.12: 108518692-108518692 on Build GRCh38
rs77241670 Chr.12: 108518622-108518622 on Build GRCh38
rs756711226 Chr.12: 108518520-108518521 on Build GRCh38
rs745970078 Chr.12: 108518532-108518532 on Build GRCh38
rs779230969 Chr.12: 108518581-108518581 on Build GRCh38
rs761360020 Chr.12: 108518551-108518551 on Build GRCh38
rs752715733 Chr.12: 108518530-108518530 on Build GRCh38
rs745904094 Chr.12: 108518597-108518597 on Build GRCh38
rs752364770 Chr.12: 108518663-108518663 on Build GRCh38
rs139799946 Chr.12: 108518636-108518636 on Build GRCh38
rs926237262 Chr.12: 108518637-108518637 on Build GRCh38
rs369952135 Chr.12: 108518573-108518573 on Build GRCh38
rs34353718 Chr.12: 108518527-108518527 on Build GRCh38
rs772531714 Chr.12: 108518558-108518558 on Build GRCh38
rs748412077 Chr.12: 108518722-108518722 on Build GRCh38
rs756923197 Chr.12: 108518701-108518701 on Build GRCh38
rs554616428 Chr.12: 108518575-108518575 on Build GRCh38
rs921333737 Chr.12: 108518679-108518679 on Build GRCh38
rs758689477 Chr.12: 108518699-108518699 on Build GRCh38
rs780151017 Chr.12: 108518612-108518612 on Build GRCh38
rs780144146 Chr.12: 108518521-108518521 on Build GRCh38
rs778841180 Chr.12: 108518531-108518531 on Build GRCh38
rs375607026 Chr.12: 108518672-108518672 on Build GRCh38
rs774068210 Chr.12: 108518643-108518643 on Build GRCh38
rs186237475 Chr.12: 108518574-108518574 on Build GRCh38
rs772940478 Chr.12: 108518638-108518638 on Build GRCh38
rs377160956 Chr.12: 108518523-108518523 on Build GRCh38

View Full Product Details