Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3660957-3661204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFRA4
Entrez Gene ID
64096
Gene Name
GDNF family receptor alpha 4
Gene Aliases
-
UniGene
Hs.302025
Gene Chromosome Location
Chr.20: 3659292-3663399 on Build GRCh38
Transcripts
NM_022139.3 NM_145762.2 XM_005260793.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5688518 c.245C>A p.A82E Chr.20: 3661091-3661091 on Build GRCh38
5343583 c.330C>T p.P110P Chr.20: 3661006-3661006 on Build GRCh38
394727 c.197G>A p.R66Q Chr.20: 3661139-3661139 on Build GRCh38
7295362 c.226C>T p.P76S Chr.20: 3661110-3661110 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778118490 Chr.20: 3661181-3661181 on Build GRCh38
rs1022128167 Chr.20: 3661048-3661048 on Build GRCh38
rs868210232 Chr.20: 3661004-3661004 on Build GRCh38
rs913989720 Chr.20: 3661041-3661041 on Build GRCh38
rs75086211 Chr.20: 3660981-3660981 on Build GRCh38
rs985463913 Chr.20: 3661121-3661121 on Build GRCh38
rs1011107107 Chr.20: 3661000-3661000 on Build GRCh38
rs930149321 Chr.20: 3661149-3661149 on Build GRCh38
rs562272020 Chr.20: 3661053-3661053 on Build GRCh38
rs943616306 Chr.20: 3661105-3661105 on Build GRCh38
rs966971276 Chr.20: 3661061-3661061 on Build GRCh38
rs529611778 Chr.20: 3661176-3661176 on Build GRCh38
rs866074771 Chr.20: 3660992-3660992 on Build GRCh38
rs756053371 Chr.20: 3661011-3661011 on Build GRCh38
rs1037940365 Chr.20: 3661119-3661119 on Build GRCh38
rs910770866 Chr.20: 3661081-3661081 on Build GRCh38
rs974585411 Chr.20: 3661134-3661134 on Build GRCh38
rs759187147 Chr.20: 3661034-3661034 on Build GRCh38
rs750234198 Chr.20: 3660976-3660976 on Build GRCh38
rs758589935 Chr.20: 3661155-3661155 on Build GRCh38
rs929477291 Chr.20: 3661139-3661139 on Build GRCh38
rs988231311 Chr.20: 3660997-3660997 on Build GRCh38
rs954711399 Chr.20: 3661063-3661063 on Build GRCh38
rs780158523 Chr.20: 3661087-3661087 on Build GRCh38
rs77187490 Chr.20: 3661075-3661075 on Build GRCh38
rs920750506 Chr.20: 3661127-3661127 on Build GRCh38
rs752785081 Chr.20: 3661115-3661115 on Build GRCh38
rs911258267 Chr.20: 3661123-3661123 on Build GRCh38
rs75836656 Chr.20: 3661028-3661028 on Build GRCh38
rs1029600676 Chr.20: 3661069-3661069 on Build GRCh38
rs764455133 Chr.20: 3661151-3661151 on Build GRCh38

View Full Product Details