Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 69171794-69172059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD4B
Entrez Gene ID
23150
Gene Name
FERM domain containing 4B
Gene Aliases
6030440G05Rik, GRSP1
UniGene
Hs.371681
Gene Chromosome Location
Chr.3: 69168782-69541340 on Build GRCh38
Transcripts
NM_015123.2 XM_005264722.1 XM_005264723.1 XM_017005989.1 XM_017005990.1 XM_017005991.1 XM_017005992.1 XM_017005993.1 XM_017005994.1 XM_017005995.1 XM_017005996.1 XM_017005997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5653471 c.3100G>C p.V1034L Chr.3: 69171866-69171866 on Build GRCh38
4405508 c.3080A>G p.D1027G Chr.3: 69171886-69171886 on Build GRCh38
4584618 c.3041A>G p.N1014S Chr.3: 69171925-69171925 on Build GRCh38
6886087 c.2902C>T p.L968F Chr.3: 69171902-69171902 on Build GRCh38
6886086 c.3064C>T p.L1022F Chr.3: 69171902-69171902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779870401 Chr.3: 69171915-69171915 on Build GRCh38
rs573865880 Chr.3: 69171830-69171830 on Build GRCh38
rs746404136 Chr.3: 69172008-69172008 on Build GRCh38
rs773187186 Chr.3: 69171832-69171832 on Build GRCh38
rs571281065 Chr.3: 69171849-69171849 on Build GRCh38
rs777417124 Chr.3: 69172026-69172026 on Build GRCh38
rs765711424 Chr.3: 69171959-69171959 on Build GRCh38
rs766147684 Chr.3: 69172008-69172008 on Build GRCh38
rs758021869 Chr.3: 69172020-69172020 on Build GRCh38
rs370422853 Chr.3: 69171929-69171929 on Build GRCh38
rs187099844 Chr.3: 69171925-69171925 on Build GRCh38
rs536092516 Chr.3: 69172030-69172030 on Build GRCh38
rs773739478 Chr.3: 69171839-69171839 on Build GRCh38
rs371111532 Chr.3: 69171829-69171829 on Build GRCh38
rs764141051 Chr.3: 69171971-69171971 on Build GRCh38
rs748073237 Chr.3: 69172019-69172019 on Build GRCh38
rs777039046 Chr.3: 69171953-69171953 on Build GRCh38
rs368186652 Chr.3: 69172010-69172010 on Build GRCh38
rs771265530 Chr.3: 69171951-69171951 on Build GRCh38
rs771781564 Chr.3: 69171836-69171836 on Build GRCh38
rs375814018 Chr.3: 69171928-69171928 on Build GRCh38
rs570143625 Chr.3: 69171993-69171993 on Build GRCh38
rs760082050 Chr.3: 69171873-69171873 on Build GRCh38
rs751823751 Chr.3: 69171984-69171984 on Build GRCh38
rs374013656 Chr.3: 69171966-69171966 on Build GRCh38
rs984611694 Chr.3: 69171858-69171858 on Build GRCh38
rs755915765 Chr.3: 69171912-69171912 on Build GRCh38
rs557068110 Chr.3: 69171964-69171964 on Build GRCh38
rs372062502 Chr.3: 69172014-69172014 on Build GRCh38
rs761068147 Chr.3: 69171843-69171843 on Build GRCh38
rs752717550 Chr.3: 69171885-69171885 on Build GRCh38
rs765289548 Chr.3: 69171875-69171875 on Build GRCh38
rs762973406 Chr.3: 69171968-69171968 on Build GRCh38
rs759976457 Chr.3: 69171958-69171958 on Build GRCh38
rs373666395 Chr.3: 69171924-69171924 on Build GRCh38
rs746628491 Chr.3: 69171937-69171937 on Build GRCh38
rs748602845 Chr.3: 69171948-69171948 on Build GRCh38
rs749254316 Chr.3: 69171922-69171922 on Build GRCh38
rs1004997350 Chr.3: 69171999-69171999 on Build GRCh38
rs773047886 Chr.3: 69171935-69171935 on Build GRCh38
rs758625950 Chr.3: 69171892-69171892 on Build GRCh38
rs761568833 Chr.3: 69171834-69171834 on Build GRCh38
rs754342263 Chr.3: 69171866-69171866 on Build GRCh38

View Full Product Details