Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27493891-27494162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC4
Entrez Gene ID
64838
Gene Name
fibronectin type III domain containing 4
Gene Aliases
FRCP1
UniGene
Hs.27836
Gene Chromosome Location
Chr.2: 27491883-27495303 on Build GRCh38
Transcripts
NM_022823.2 XM_005264499.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1019724 c.251G>A p.R84Q Chr.2: 27494133-27494133 on Build GRCh38
1019723 c.407G>A p.R136Q Chr.2: 27493977-27493977 on Build GRCh38
7241918 c.378G>C p.E126D Chr.2: 27494006-27494006 on Build GRCh38
7258646 c.356G>T p.R119M Chr.2: 27494028-27494028 on Build GRCh38
7087360 c.378G>A p.E126E Chr.2: 27494006-27494006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755064284 Chr.2: 27494101-27494101 on Build GRCh38
rs200150285 Chr.2: 27493931-27493931 on Build GRCh38
rs773717899 Chr.2: 27494083-27494083 on Build GRCh38
rs759239659 Chr.2: 27493957-27493957 on Build GRCh38
rs774100586 Chr.2: 27493942-27493942 on Build GRCh38
rs999619159 Chr.2: 27494094-27494094 on Build GRCh38
rs1039933406 Chr.2: 27493925-27493925 on Build GRCh38
rs771833073 Chr.2: 27494035-27494035 on Build GRCh38
rs765717697 Chr.2: 27493967-27493967 on Build GRCh38
rs779169265 Chr.2: 27494033-27494033 on Build GRCh38
rs977458631 Chr.2: 27493956-27493956 on Build GRCh38
rs751800976 Chr.2: 27494011-27494012 on Build GRCh38
rs758463099 Chr.2: 27493985-27493985 on Build GRCh38
rs777730572 Chr.2: 27494014-27494014 on Build GRCh38
rs777045269 Chr.2: 27493960-27493960 on Build GRCh38
rs762170194 Chr.2: 27494045-27494045 on Build GRCh38
rs766467845 Chr.2: 27493990-27493990 on Build GRCh38
rs765550102 Chr.2: 27494076-27494076 on Build GRCh38
rs773966063 Chr.2: 27494025-27494025 on Build GRCh38
rs942511052 Chr.2: 27493914-27493914 on Build GRCh38
rs144942318 Chr.2: 27493937-27493937 on Build GRCh38
rs780934969 Chr.2: 27493913-27493913 on Build GRCh38
rs1046558 Chr.2: 27494020-27494020 on Build GRCh38
rs755893016 Chr.2: 27494120-27494120 on Build GRCh38
rs766940325 Chr.2: 27494091-27494091 on Build GRCh38
rs777410525 Chr.2: 27494127-27494127 on Build GRCh38
rs200705101 Chr.2: 27493994-27493994 on Build GRCh38
rs369929899 Chr.2: 27494138-27494138 on Build GRCh38
rs545905554 Chr.2: 27494104-27494104 on Build GRCh38
rs902486387 Chr.2: 27494084-27494084 on Build GRCh38
rs762138570 Chr.2: 27493961-27493961 on Build GRCh38
rs767315126 Chr.2: 27493958-27493958 on Build GRCh38
rs781451528 Chr.2: 27494010-27494010 on Build GRCh38
rs780049304 Chr.2: 27494003-27494004 on Build GRCh38
rs938775656 Chr.2: 27493978-27493978 on Build GRCh38
rs542003816 Chr.2: 27493991-27493991 on Build GRCh38
rs752898033 Chr.2: 27494111-27494111 on Build GRCh38
rs751625552 Chr.2: 27494098-27494098 on Build GRCh38
rs755186709 Chr.2: 27494083-27494083 on Build GRCh38
rs138262822 Chr.2: 27494036-27494036 on Build GRCh38
rs747960631 Chr.2: 27493922-27493922 on Build GRCh38
rs747919954 Chr.2: 27494013-27494013 on Build GRCh38
rs563276232 Chr.2: 27493954-27493954 on Build GRCh38
rs371859539 Chr.2: 27494141-27494141 on Build GRCh38
rs750897858 Chr.2: 27493970-27493970 on Build GRCh38
rs763421619 Chr.2: 27494086-27494086 on Build GRCh38
rs61734464 Chr.2: 27494021-27494021 on Build GRCh38
rs555149490 Chr.2: 27493926-27493926 on Build GRCh38

View Full Product Details