Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89186302-89186565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP4
Entrez Gene ID
115361
Gene Name
guanylate binding protein 4
Gene Aliases
Mpa2
UniGene
Hs.409925
Gene Chromosome Location
Chr.1: 89181148-89198950 on Build GRCh38
Transcripts
NM_052941.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669799 c.1707+1G>A p.? Chr.1: 89186332-89186332 on Build GRCh38
2258908 c.1648G>A p.E550K Chr.1: 89186392-89186392 on Build GRCh38
2258909 c.1569delA p.K523fs*8 Chr.1: 89186471-89186471 on Build GRCh38
6127924 c.1527G>A p.M509I Chr.1: 89186513-89186513 on Build GRCh38
358159 c.1515G>A p.A505A Chr.1: 89186525-89186525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769580903 Chr.1: 89186490-89186490 on Build GRCh38
rs190249684 Chr.1: 89186430-89186430 on Build GRCh38
rs655260 Chr.1: 89186419-89186419 on Build GRCh38
rs1142886 Chr.1: 89186414-89186414 on Build GRCh38
rs754127744 Chr.1: 89186418-89186418 on Build GRCh38
rs561042 Chr.1: 89186388-89186388 on Build GRCh38
rs557859571 Chr.1: 89186326-89186326 on Build GRCh38
rs772735640 Chr.1: 89186365-89186365 on Build GRCh38
rs1142889 Chr.1: 89186405-89186405 on Build GRCh38
rs773408251 Chr.1: 89186517-89186517 on Build GRCh38
rs764190754 Chr.1: 89186516-89186516 on Build GRCh38
rs386633304 Chr.1: 89186404-89186407 on Build GRCh38
rs746588630 Chr.1: 89186451-89186451 on Build GRCh38
rs377205873 Chr.1: 89186323-89186323 on Build GRCh38
rs904895437 Chr.1: 89186519-89186519 on Build GRCh38
rs771533053 Chr.1: 89186355-89186355 on Build GRCh38
rs1142887 Chr.1: 89186411-89186411 on Build GRCh38
rs1142890 Chr.1: 89186404-89186404 on Build GRCh38
rs386633303 Chr.1: 89186388-89186389 on Build GRCh38
rs776395231 Chr.1: 89186484-89186484 on Build GRCh38
rs188809753 Chr.1: 89186521-89186521 on Build GRCh38
rs1142888 Chr.1: 89186407-89186407 on Build GRCh38
rs551457653 Chr.1: 89186525-89186525 on Build GRCh38
rs760265579 Chr.1: 89186379-89186379 on Build GRCh38
rs770559828 Chr.1: 89186461-89186461 on Build GRCh38
rs765064674 Chr.1: 89186396-89186396 on Build GRCh38
rs529438124 Chr.1: 89186446-89186446 on Build GRCh38
rs746860813 Chr.1: 89186360-89186360 on Build GRCh38
rs148706291 Chr.1: 89186420-89186420 on Build GRCh38
rs151234239 Chr.1: 89186485-89186485 on Build GRCh38
rs879722090 Chr.1: 89186404-89186405 on Build GRCh38
rs777198893 Chr.1: 89186450-89186450 on Build GRCh38
rs144257890 Chr.1: 89186399-89186399 on Build GRCh38
rs961119368 Chr.1: 89186528-89186528 on Build GRCh38
rs146992170 Chr.1: 89186520-89186520 on Build GRCh38
rs201719385 Chr.1: 89186350-89186350 on Build GRCh38
rs762915965 Chr.1: 89186510-89186510 on Build GRCh38
rs1035583714 Chr.1: 89186338-89186338 on Build GRCh38
rs879604683 Chr.1: 89186388-89186389 on Build GRCh38
rs142011957 Chr.1: 89186387-89186387 on Build GRCh38
rs775161494 Chr.1: 89186509-89186509 on Build GRCh38
rs747584155 Chr.1: 89186349-89186349 on Build GRCh38
rs994751233 Chr.1: 89186500-89186500 on Build GRCh38
rs776683474 Chr.1: 89186453-89186454 on Build GRCh38
rs932718230 Chr.1: 89186394-89186394 on Build GRCh38
rs373601177 Chr.1: 89186322-89186322 on Build GRCh38
rs140981637 Chr.1: 89186478-89186478 on Build GRCh38
rs561037 Chr.1: 89186389-89186389 on Build GRCh38
rs748405713 Chr.1: 89186475-89186476 on Build GRCh38
rs768779188 Chr.1: 89186396-89186398 on Build GRCh38
rs608339 Chr.1: 89186395-89186395 on Build GRCh38
rs368648285 Chr.1: 89186526-89186526 on Build GRCh38

View Full Product Details