Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10768944-10769217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUBP1
Entrez Gene ID
4682
Gene Name
nucleotide binding protein 1
Gene Aliases
NBP, NBP1, NBP35
UniGene
Hs.81469
Gene Chromosome Location
Chr.16: 10743164-10769351 on Build GRCh38
Transcripts
NM_001278506.1 NM_001323594.1 NM_001323595.1 NM_001323596.1 NM_001323597.1 NM_002484.3 XM_017023252.1 NM_001079512.3 NM_001318873.1 NR_134901.1 NR_134902.1 NR_134903.1 NR_134904.1 NR_134905.1 NR_134906.1 XM_006720944.3 XM_011522654.2 XM_017023649.1 XM_017023650.1 XM_017023651.1

Gene Symbol
TVP23A
Entrez Gene ID
780776
Gene Name
trans-golgi network vesicle protein 23 homolog A
Gene Aliases
FAM18A, YDR084C
UniGene
Hs.371576
Gene Chromosome Location
Chr.16: 10758034-10818902 on Build GRCh38
Transcripts
NM_001278506.1 NM_001323594.1 NM_001323595.1 NM_001323596.1 NM_001323597.1 NM_002484.3 XM_017023252.1 NM_001079512.3 NM_001318873.1 NR_134901.1 NR_134902.1 NR_134903.1 NR_134904.1 NR_134905.1 NR_134906.1 XM_006720944.3 XM_011522654.2 XM_017023649.1 XM_017023650.1 XM_017023651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6307862 c.957T>G p.S319R Chr.16: 10769099-10769099 on Build GRCh38
6580128 c.935C>G p.S312* Chr.16: 10769077-10769077 on Build GRCh38
7380649 c.954C>G p.I318M Chr.16: 10769096-10769096 on Build GRCh38
966936 c.945G>T p.E315D Chr.16: 10769087-10769087 on Build GRCh38
5473674 c.950T>C p.L317P Chr.16: 10769092-10769092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776450811 Chr.16: 10769081-10769081 on Build GRCh38
rs906355039 Chr.16: 10769178-10769178 on Build GRCh38
rs190573534 Chr.16: 10769192-10769192 on Build GRCh38
rs200227375 Chr.16: 10769097-10769097 on Build GRCh38
rs978697762 Chr.16: 10768998-10768998 on Build GRCh38
rs766203356 Chr.16: 10769007-10769007 on Build GRCh38
rs774868931 Chr.16: 10769174-10769174 on Build GRCh38
rs778997931 Chr.16: 10769150-10769150 on Build GRCh38
rs550291329 Chr.16: 10769191-10769191 on Build GRCh38
rs757236292 Chr.16: 10769142-10769142 on Build GRCh38
rs201978663 Chr.16: 10769020-10769020 on Build GRCh38
rs1002389242 Chr.16: 10769181-10769181 on Build GRCh38
rs2233542 Chr.16: 10769022-10769022 on Build GRCh38
rs1039207477 Chr.16: 10769109-10769109 on Build GRCh38
rs369062540 Chr.16: 10769029-10769029 on Build GRCh38
rs923003592 Chr.16: 10769116-10769116 on Build GRCh38
rs533400522 Chr.16: 10769189-10769189 on Build GRCh38
rs746112256 Chr.16: 10769074-10769074 on Build GRCh38
rs386788939 Chr.16: 10769107-10769108 on Build GRCh38
rs920620743 Chr.16: 10769130-10769130 on Build GRCh38
rs11716 Chr.16: 10769107-10769107 on Build GRCh38
rs757360350 Chr.16: 10769056-10769056 on Build GRCh38
rs987567110 Chr.16: 10769071-10769071 on Build GRCh38
rs749394368 Chr.16: 10769049-10769049 on Build GRCh38
rs879057877 Chr.16: 10769155-10769155 on Build GRCh38
rs779047393 Chr.16: 10769062-10769062 on Build GRCh38
rs768402371 Chr.16: 10769075-10769075 on Build GRCh38
rs1025036731 Chr.16: 10769195-10769195 on Build GRCh38
rs1004019705 Chr.16: 10768983-10768983 on Build GRCh38
rs756625301 Chr.16: 10769030-10769030 on Build GRCh38
rs939837291 Chr.16: 10769176-10769176 on Build GRCh38
rs185648760 Chr.16: 10769137-10769137 on Build GRCh38
rs1046300600 Chr.16: 10769177-10769177 on Build GRCh38
rs970425281 Chr.16: 10768997-10768997 on Build GRCh38
rs564302332 Chr.16: 10769141-10769141 on Build GRCh38
rs2233543 Chr.16: 10769108-10769108 on Build GRCh38
rs146511825 Chr.16: 10769193-10769193 on Build GRCh38
rs1024737244 Chr.16: 10768991-10768991 on Build GRCh38
rs777935565 Chr.16: 10769069-10769070 on Build GRCh38
rs527993353 Chr.16: 10769111-10769111 on Build GRCh38
rs201618624 Chr.16: 10769033-10769033 on Build GRCh38
rs772778325 Chr.16: 10769104-10769104 on Build GRCh38
rs767967750 Chr.16: 10769125-10769125 on Build GRCh38
rs748096118 Chr.16: 10769084-10769084 on Build GRCh38
rs376901396 Chr.16: 10769138-10769138 on Build GRCh38

View Full Product Details