Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190881304-190881575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLS
Entrez Gene ID
2744
Gene Name
glutaminase
Gene Aliases
AAD20, GAC, GAM, GLS1, KGA
UniGene
Hs.116448
Gene Chromosome Location
Chr.2: 190880821-190965552 on Build GRCh38
Transcripts
NM_001256310.1 NM_014905.4 XM_005246467.3 XM_006712435.1 XM_017003829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4139142 c.246G>A p.E82E Chr.2: 190881330-190881330 on Build GRCh38
5993731 c.377C>A p.S126* Chr.2: 190881461-190881461 on Build GRCh38
5993732 c.377C>A p.S126* Chr.2: 190881461-190881461 on Build GRCh38
4139141 c.246G>A p.E82E Chr.2: 190881330-190881330 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768903785 Chr.2: 190881331-190881331 on Build GRCh38
rs965841032 Chr.2: 190881397-190881397 on Build GRCh38
rs527974778 Chr.2: 190881339-190881339 on Build GRCh38
rs889058089 Chr.2: 190881370-190881370 on Build GRCh38
rs780073821 Chr.2: 190881497-190881497 on Build GRCh38
rs1018309361 Chr.2: 190881378-190881378 on Build GRCh38
rs1037520347 Chr.2: 190881494-190881494 on Build GRCh38
rs757622923 Chr.2: 190881550-190881550 on Build GRCh38
rs939066682 Chr.2: 190881517-190881517 on Build GRCh38
rs763896449 Chr.2: 190881451-190881451 on Build GRCh38
rs979379662 Chr.2: 190881543-190881543 on Build GRCh38
rs773785583 Chr.2: 190881393-190881393 on Build GRCh38
rs994744051 Chr.2: 190881362-190881362 on Build GRCh38
rs537971593 Chr.2: 190881374-190881374 on Build GRCh38
rs764871516 Chr.2: 190881427-190881427 on Build GRCh38
rs374890296 Chr.2: 190881493-190881493 on Build GRCh38
rs934925718 Chr.2: 190881349-190881349 on Build GRCh38
rs932725310 Chr.2: 190881444-190881444 on Build GRCh38
rs751307120 Chr.2: 190881459-190881459 on Build GRCh38
rs768864719 Chr.2: 190881505-190881505 on Build GRCh38
rs956496560 Chr.2: 190881546-190881546 on Build GRCh38
rs758922808 Chr.2: 190881394-190881394 on Build GRCh38
rs780929751 Chr.2: 190881485-190881485 on Build GRCh38
rs897687240 Chr.2: 190881357-190881357 on Build GRCh38
rs998248129 Chr.2: 190881406-190881406 on Build GRCh38
rs78219026 Chr.2: 190881507-190881507 on Build GRCh38
rs550215381 Chr.2: 190881434-190881434 on Build GRCh38
rs993395684 Chr.2: 190881504-190881504 on Build GRCh38
rs1036143990 Chr.2: 190881525-190881525 on Build GRCh38
rs1022698166 Chr.2: 190881524-190881524 on Build GRCh38
rs538979300 Chr.2: 190881496-190881496 on Build GRCh38
rs758103656 Chr.2: 190881448-190881448 on Build GRCh38
rs752297037 Chr.2: 190881432-190881432 on Build GRCh38
rs767849917 Chr.2: 190881389-190881389 on Build GRCh38
rs377700631 Chr.2: 190881518-190881518 on Build GRCh38
rs1031114903 Chr.2: 190881410-190881410 on Build GRCh38
rs895464953 Chr.2: 190881511-190881511 on Build GRCh38
rs1049814856 Chr.2: 190881462-190881462 on Build GRCh38
rs757078343 Chr.2: 190881484-190881484 on Build GRCh38
rs549201657 Chr.2: 190881340-190881340 on Build GRCh38
rs1048890021 Chr.2: 190881548-190881548 on Build GRCh38
rs147276225 Chr.2: 190881502-190881502 on Build GRCh38
rs778237907 Chr.2: 190881506-190881506 on Build GRCh38
rs1049421722 Chr.2: 190881369-190881369 on Build GRCh38
rs968421988 Chr.2: 190881536-190881536 on Build GRCh38
rs1031830276 Chr.2: 190881544-190881544 on Build GRCh38

View Full Product Details