Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45711908-45712167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO46
Entrez Gene ID
23403
Gene Name
F-box protein 46
Gene Aliases
20D7-FC4, FBXO34L, Fbx46
UniGene
Hs.128702
Gene Chromosome Location
Chr.19: 45710629-45730893 on Build GRCh38
Transcripts
NM_001080469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
118329 c.1509C>T p.F503F Chr.19: 45711987-45711987 on Build GRCh38
6662346 c.1473C>T p.R491R Chr.19: 45712023-45712023 on Build GRCh38
4581222 c.1576C>A p.R526S Chr.19: 45711920-45711920 on Build GRCh38
4079433 c.1476G>A p.A492A Chr.19: 45712020-45712020 on Build GRCh38
6895258 c.1582G>T p.D528Y Chr.19: 45711914-45711914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755064999 Chr.19: 45712061-45712061 on Build GRCh38
rs763244125 Chr.19: 45712039-45712039 on Build GRCh38
rs370707906 Chr.19: 45712026-45712026 on Build GRCh38
rs757208473 Chr.19: 45712076-45712076 on Build GRCh38
rs749212499 Chr.19: 45711943-45711943 on Build GRCh38
rs960198972 Chr.19: 45712104-45712104 on Build GRCh38
rs745581898 Chr.19: 45712087-45712087 on Build GRCh38
rs778807365 Chr.19: 45711959-45711959 on Build GRCh38
rs935610559 Chr.19: 45712002-45712002 on Build GRCh38
rs757155522 Chr.19: 45711951-45711951 on Build GRCh38
rs758208859 Chr.19: 45712101-45712101 on Build GRCh38
rs746649463 Chr.19: 45712134-45712134 on Build GRCh38
rs1052373589 Chr.19: 45712012-45712012 on Build GRCh38
rs753855427 Chr.19: 45712065-45712065 on Build GRCh38
rs779629112 Chr.19: 45712125-45712125 on Build GRCh38
rs770220571 Chr.19: 45712147-45712147 on Build GRCh38
rs777919168 Chr.19: 45711930-45711930 on Build GRCh38
rs746566264 Chr.19: 45712001-45712001 on Build GRCh38
rs766581156 Chr.19: 45712041-45712041 on Build GRCh38
rs192506598 Chr.19: 45712137-45712137 on Build GRCh38
rs745553569 Chr.19: 45711963-45711963 on Build GRCh38
rs780848834 Chr.19: 45712027-45712027 on Build GRCh38
rs976598153 Chr.19: 45712113-45712113 on Build GRCh38
rs61757392 Chr.19: 45712014-45712014 on Build GRCh38
rs767670856 Chr.19: 45712062-45712062 on Build GRCh38
rs376057661 Chr.19: 45711991-45711991 on Build GRCh38
rs751795632 Chr.19: 45712047-45712047 on Build GRCh38
rs868002289 Chr.19: 45712096-45712097 on Build GRCh38
rs555629800 Chr.19: 45712143-45712143 on Build GRCh38
rs771566097 Chr.19: 45711983-45711983 on Build GRCh38
rs966143894 Chr.19: 45712070-45712070 on Build GRCh38
rs768197332 Chr.19: 45712011-45712011 on Build GRCh38
rs1018225975 Chr.19: 45712023-45712023 on Build GRCh38

View Full Product Details