Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30498710-30498969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGCT
Entrez Gene ID
79017
Gene Name
gamma-glutamylcyclotransferase
Gene Aliases
C7orf24, CRF21, GCTG, GGC
UniGene
Hs.530024
Gene Chromosome Location
Chr.7: 30496621-30504841 on Build GRCh38
Transcripts
NM_001199815.1 NM_001199816.1 NM_001199817.1 NM_024051.3 NR_037669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1450337 c.338C>A p.A113E Chr.7: 30498888-30498888 on Build GRCh38
6471860 c.378G>A p.L126L Chr.7: 30498848-30498848 on Build GRCh38
452956 c.390C>G p.Y130* Chr.7: 30498836-30498836 on Build GRCh38
1622775 c.367C>G p.R123G Chr.7: 30498859-30498859 on Build GRCh38
6565555 c.294A>T p.E98D Chr.7: 30498932-30498932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775341582 Chr.7: 30498813-30498813 on Build GRCh38
rs142543540 Chr.7: 30498826-30498826 on Build GRCh38
rs749023003 Chr.7: 30498905-30498905 on Build GRCh38
rs755625526 Chr.7: 30498757-30498758 on Build GRCh38
rs200366394 Chr.7: 30498790-30498790 on Build GRCh38
rs367645393 Chr.7: 30498760-30498760 on Build GRCh38
rs753110206 Chr.7: 30498836-30498836 on Build GRCh38
rs769526034 Chr.7: 30498802-30498802 on Build GRCh38
rs534171650 Chr.7: 30498941-30498941 on Build GRCh38
rs758964744 Chr.7: 30498773-30498773 on Build GRCh38
rs757316854 Chr.7: 30498883-30498883 on Build GRCh38
rs953940798 Chr.7: 30498825-30498825 on Build GRCh38
rs917184898 Chr.7: 30498844-30498844 on Build GRCh38
rs747020968 Chr.7: 30498778-30498778 on Build GRCh38
rs375218724 Chr.7: 30498921-30498921 on Build GRCh38
rs13238986 Chr.7: 30498894-30498894 on Build GRCh38
rs746068169 Chr.7: 30498798-30498798 on Build GRCh38
rs779855606 Chr.7: 30498865-30498868 on Build GRCh38
rs760357690 Chr.7: 30498834-30498834 on Build GRCh38
rs758508499 Chr.7: 30498859-30498859 on Build GRCh38
rs758981424 Chr.7: 30498849-30498849 on Build GRCh38
rs762942989 Chr.7: 30498818-30498818 on Build GRCh38
rs779826560 Chr.7: 30498903-30498903 on Build GRCh38
rs756354974 Chr.7: 30498900-30498900 on Build GRCh38
rs778067732 Chr.7: 30498774-30498774 on Build GRCh38
rs753308737 Chr.7: 30498770-30498770 on Build GRCh38
rs781305842 Chr.7: 30498885-30498885 on Build GRCh38
rs1039430154 Chr.7: 30498919-30498919 on Build GRCh38
rs746133607 Chr.7: 30498899-30498899 on Build GRCh38
rs748047628 Chr.7: 30498929-30498929 on Build GRCh38
rs767231584 Chr.7: 30498831-30498831 on Build GRCh38
rs71532977 Chr.7: 30498783-30498783 on Build GRCh38
rs371796444 Chr.7: 30498858-30498858 on Build GRCh38
rs765582824 Chr.7: 30498835-30498835 on Build GRCh38
rs760722305 Chr.7: 30498945-30498945 on Build GRCh38
rs13238987 Chr.7: 30498895-30498895 on Build GRCh38

View Full Product Details