Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76540404-76540635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYGB
Entrez Gene ID
114757
Gene Name
cytoglobin
Gene Aliases
HGB, STAP
UniGene
Hs.95120
Gene Chromosome Location
Chr.17: 76527348-76557692 on Build GRCh38
Transcripts
XM_017024116.1 NM_001077620.2 NR_033357.1 XM_017025013.1 XM_017025014.1 XM_017025015.1

Gene Symbol
PRCD
Entrez Gene ID
768206
Gene Name
progressive rod-cone degeneration
Gene Aliases
RP36
UniGene
Hs.634380
Gene Chromosome Location
Chr.17: 76527586-76555339 on Build GRCh38
Transcripts
XM_017024116.1 NM_001077620.2 NR_033357.1 XM_017025013.1 XM_017025014.1 XM_017025015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1221864 c.107G>A p.G36D Chr.17: 76540537-76540537 on Build GRCh38
7321584 c.84C>T p.S28S Chr.17: 76540514-76540514 on Build GRCh38
4563351 c.96G>A p.G32G Chr.17: 76540526-76540526 on Build GRCh38
5464208 c.143+4A>C p.? Chr.17: 76540577-76540577 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778402370 Chr.17: 76540590-76540590 on Build GRCh38
rs751944355 Chr.17: 76540464-76540464 on Build GRCh38
rs753270253 Chr.17: 76540540-76540540 on Build GRCh38
rs200091367 Chr.17: 76540502-76540502 on Build GRCh38
rs770202626 Chr.17: 76540588-76540588 on Build GRCh38
rs750104756 Chr.17: 76540553-76540553 on Build GRCh38
rs778591141 Chr.17: 76540472-76540472 on Build GRCh38
rs372084413 Chr.17: 76540515-76540515 on Build GRCh38
rs760532713 Chr.17: 76540529-76540529 on Build GRCh38
rs368314608 Chr.17: 76540539-76540539 on Build GRCh38
rs781553592 Chr.17: 76540556-76540556 on Build GRCh38
rs374419323 Chr.17: 76540525-76540525 on Build GRCh38
rs528833785 Chr.17: 76540469-76540469 on Build GRCh38
rs775143019 Chr.17: 76540616-76540616 on Build GRCh38
rs975797148 Chr.17: 76540589-76540589 on Build GRCh38
rs764158636 Chr.17: 76540534-76540534 on Build GRCh38
rs753462939 Chr.17: 76540465-76540465 on Build GRCh38
rs748628692 Chr.17: 76540477-76540477 on Build GRCh38
rs767439982 Chr.17: 76540518-76540518 on Build GRCh38
rs769857895 Chr.17: 76540481-76540481 on Build GRCh38
rs867338006 Chr.17: 76540552-76540552 on Build GRCh38
rs759376499 Chr.17: 76540517-76540517 on Build GRCh38
rs749307328 Chr.17: 76540593-76540593 on Build GRCh38
rs778503433 Chr.17: 76540548-76540548 on Build GRCh38
rs771924891 Chr.17: 76540609-76540609 on Build GRCh38
rs763273515 Chr.17: 76540426-76540426 on Build GRCh38
rs1009828411 Chr.17: 76540524-76540533 on Build GRCh38
rs886053474 Chr.17: 76540495-76540495 on Build GRCh38
rs1023722325 Chr.17: 76540562-76540562 on Build GRCh38
rs769046801 Chr.17: 76540476-76540476 on Build GRCh38
rs1051400643 Chr.17: 76540439-76540439 on Build GRCh38
rs771176701 Chr.17: 76540594-76540594 on Build GRCh38
rs747350100 Chr.17: 76540475-76540475 on Build GRCh38
rs752832187 Chr.17: 76540616-76540617 on Build GRCh38
rs781509601 Chr.17: 76540614-76540615 on Build GRCh38
rs568895462 Chr.17: 76540547-76540547 on Build GRCh38
rs759849474 Chr.17: 76540604-76540604 on Build GRCh38
rs368685571 Chr.17: 76540442-76540442 on Build GRCh38
rs748625799 Chr.17: 76540564-76540564 on Build GRCh38
rs755599869 Chr.17: 76540461-76540461 on Build GRCh38

View Full Product Details