Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89116867-89117134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP2
Entrez Gene ID
2634
Gene Name
guanylate binding protein 2
Gene Aliases
-
UniGene
Hs.386567
Gene Chromosome Location
Chr.1: 89106132-89126159 on Build GRCh38
Transcripts
NM_004120.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4686544 c.836C>T p.T279I Chr.1: 89117024-89117024 on Build GRCh38
146189 c.868+1G>C p.? Chr.1: 89116991-89116991 on Build GRCh38
5922020 c.868+4A>G p.? Chr.1: 89116988-89116988 on Build GRCh38
4010438 c.809A>G p.Y270C Chr.1: 89117051-89117051 on Build GRCh38
4808798 c.814C>G p.L272V Chr.1: 89117046-89117046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2230336 Chr.1: 89117019-89117019 on Build GRCh38
rs888847270 Chr.1: 89117054-89117054 on Build GRCh38
rs769514280 Chr.1: 89117024-89117024 on Build GRCh38
rs756205791 Chr.1: 89116973-89116973 on Build GRCh38
rs778174916 Chr.1: 89116968-89116968 on Build GRCh38
rs751517507 Chr.1: 89116970-89116970 on Build GRCh38
rs765372923 Chr.1: 89117079-89117079 on Build GRCh38
rs778042009 Chr.1: 89117106-89117106 on Build GRCh38
rs766417117 Chr.1: 89116976-89116976 on Build GRCh38
rs763853855 Chr.1: 89117033-89117033 on Build GRCh38
rs762836659 Chr.1: 89117032-89117032 on Build GRCh38
rs755060182 Chr.1: 89117078-89117078 on Build GRCh38
rs748450335 Chr.1: 89117000-89117000 on Build GRCh38
rs761695136 Chr.1: 89117042-89117042 on Build GRCh38
rs754059516 Chr.1: 89116989-89116989 on Build GRCh38
rs918214740 Chr.1: 89116980-89116980 on Build GRCh38
rs968360796 Chr.1: 89117097-89117097 on Build GRCh38
rs775287224 Chr.1: 89117026-89117026 on Build GRCh38
rs764057835 Chr.1: 89116954-89116954 on Build GRCh38
rs774321393 Chr.1: 89117036-89117036 on Build GRCh38
rs370885169 Chr.1: 89117012-89117012 on Build GRCh38
rs778334754 Chr.1: 89117006-89117006 on Build GRCh38
rs758744642 Chr.1: 89117089-89117089 on Build GRCh38
rs771627137 Chr.1: 89117036-89117037 on Build GRCh38
rs777496524 Chr.1: 89116941-89116941 on Build GRCh38
rs767518146 Chr.1: 89117043-89117043 on Build GRCh38
rs142698251 Chr.1: 89117003-89117003 on Build GRCh38
rs185011739 Chr.1: 89116991-89116991 on Build GRCh38
rs762855111 Chr.1: 89116951-89116951 on Build GRCh38
rs1803632 Chr.1: 89117007-89117007 on Build GRCh38
rs753809869 Chr.1: 89117073-89117073 on Build GRCh38
rs375023862 Chr.1: 89117088-89117088 on Build GRCh38
rs1046971244 Chr.1: 89116982-89116982 on Build GRCh38
rs749738138 Chr.1: 89116991-89116992 on Build GRCh38
rs886838509 Chr.1: 89116999-89116999 on Build GRCh38
rs931131992 Chr.1: 89116907-89116907 on Build GRCh38
rs149789032 Chr.1: 89116992-89116992 on Build GRCh38
rs775965374 Chr.1: 89117014-89117014 on Build GRCh38

View Full Product Details