Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46149986-46150247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTCD
Entrez Gene ID
10841
Gene Name
formimidoyltransferase cyclodeaminase
Gene Aliases
LCHC1
UniGene
Hs.415846
Gene Chromosome Location
Chr.21: 46135981-46156481 on Build GRCh38
Transcripts
NM_001320412.1 NM_006657.2 NM_206965.1 XM_006723961.3 XM_006723962.3 XM_011529434.2 XM_011529435.2 XM_011529436.2 XM_011529437.2 XM_011529439.2 XM_011529440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4561401 c.883G>A p.E295K Chr.21: 46150142-46150142 on Build GRCh38
6094817 c.842C>T p.A281V Chr.21: 46150183-46150183 on Build GRCh38
5393082 c.897G>A p.R299R Chr.21: 46150128-46150128 on Build GRCh38
6355213 c.814C>T p.L272L Chr.21: 46150211-46150211 on Build GRCh38
6299065 c.897G>C p.R299R Chr.21: 46150128-46150128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766253469 Chr.21: 46150179-46150179 on Build GRCh38
rs878942547 Chr.21: 46150084-46150084 on Build GRCh38
rs760635509 Chr.21: 46150178-46150178 on Build GRCh38
rs376001900 Chr.21: 46150112-46150112 on Build GRCh38
rs746554639 Chr.21: 46150087-46150088 on Build GRCh38
rs368811508 Chr.21: 46150103-46150103 on Build GRCh38
rs747491063 Chr.21: 46150130-46150130 on Build GRCh38
rs530255782 Chr.21: 46150068-46150068 on Build GRCh38
rs573941266 Chr.21: 46150106-46150106 on Build GRCh38
rs566161120 Chr.21: 46150056-46150056 on Build GRCh38
rs774045878 Chr.21: 46150164-46150164 on Build GRCh38
rs752773821 Chr.21: 46150096-46150096 on Build GRCh38
rs777944418 Chr.21: 46150209-46150209 on Build GRCh38
rs544050099 Chr.21: 46150095-46150095 on Build GRCh38
rs376851412 Chr.21: 46150182-46150182 on Build GRCh38
rs759698021 Chr.21: 46150061-46150061 on Build GRCh38
rs539856624 Chr.21: 46150062-46150062 on Build GRCh38
rs534808605 Chr.21: 46150020-46150020 on Build GRCh38
rs777453453 Chr.21: 46150084-46150084 on Build GRCh38
rs777161061 Chr.21: 46150139-46150139 on Build GRCh38
rs561348258 Chr.21: 46150021-46150021 on Build GRCh38
rs761284163 Chr.21: 46150166-46150166 on Build GRCh38
rs771538518 Chr.21: 46150169-46150171 on Build GRCh38
rs576667429 Chr.21: 46150093-46150093 on Build GRCh38
rs138650765 Chr.21: 46150169-46150169 on Build GRCh38
rs374148439 Chr.21: 46150082-46150082 on Build GRCh38
rs112198665 Chr.21: 46150190-46150190 on Build GRCh38
rs375796852 Chr.21: 46150111-46150111 on Build GRCh38
rs200444548 Chr.21: 46150134-46150134 on Build GRCh38
rs768195596 Chr.21: 46150152-46150152 on Build GRCh38
rs762729365 Chr.21: 46150092-46150092 on Build GRCh38
rs1048710136 Chr.21: 46150201-46150201 on Build GRCh38
rs558104383 Chr.21: 46150066-46150066 on Build GRCh38
rs1005109558 Chr.21: 46150036-46150036 on Build GRCh38
rs76474184 Chr.21: 46150104-46150104 on Build GRCh38
rs752941882 Chr.21: 46150183-46150183 on Build GRCh38
rs767027376 Chr.21: 46150167-46150167 on Build GRCh38
rs372299678 Chr.21: 46150168-46150168 on Build GRCh38
rs755213661 Chr.21: 46150108-46150108 on Build GRCh38
rs1016205966 Chr.21: 46150042-46150042 on Build GRCh38
rs119469015 Chr.21: 46150129-46150129 on Build GRCh38

View Full Product Details