Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78094763-78095022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPC2
Entrez Gene ID
54810
Gene Name
GIPC PDZ domain containing family member 2
Gene Aliases
SEMCAP-2, SEMCAP2
UniGene
Hs.603818
Gene Chromosome Location
Chr.1: 78044962-78138449 on Build GRCh38
Transcripts
NM_001304725.1 NM_017655.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
912353 c.428G>A p.R143K Chr.1: 78094953-78094953 on Build GRCh38
912354 c.428G>T p.R143I Chr.1: 78094953-78094953 on Build GRCh38
682022 c.481C>G p.H161D Chr.1: 78095006-78095006 on Build GRCh38
244700 c.456A>G p.S152S Chr.1: 78094981-78094981 on Build GRCh38
912355 c.480T>G p.D160E Chr.1: 78095005-78095005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753881131 Chr.1: 78094916-78094916 on Build GRCh38
rs775947410 Chr.1: 78094976-78094976 on Build GRCh38
rs202235302 Chr.1: 78094929-78094929 on Build GRCh38
rs771732963 Chr.1: 78094933-78094935 on Build GRCh38
rs778621972 Chr.1: 78094926-78094926 on Build GRCh38
rs982014876 Chr.1: 78094991-78094991 on Build GRCh38
rs556578202 Chr.1: 78094989-78094989 on Build GRCh38
rs1000056492 Chr.1: 78094805-78094805 on Build GRCh38
rs766215078 Chr.1: 78094909-78094909 on Build GRCh38
rs1041671405 Chr.1: 78094796-78094797 on Build GRCh38
rs903090979 Chr.1: 78094797-78094797 on Build GRCh38
rs893929244 Chr.1: 78094836-78094836 on Build GRCh38
rs1032843606 Chr.1: 78094809-78094809 on Build GRCh38
rs763440770 Chr.1: 78094978-78094978 on Build GRCh38
rs1015158138 Chr.1: 78094953-78094953 on Build GRCh38
rs746476493 Chr.1: 78094940-78094940 on Build GRCh38
rs540210721 Chr.1: 78094961-78094961 on Build GRCh38
rs770436959 Chr.1: 78094975-78094975 on Build GRCh38
rs746315949 Chr.1: 78094974-78094974 on Build GRCh38
rs1030701945 Chr.1: 78094911-78094911 on Build GRCh38
rs774694759 Chr.1: 78094993-78094993 on Build GRCh38
rs777445472 Chr.1: 78094939-78094939 on Build GRCh38
rs754916013 Chr.1: 78094918-78094918 on Build GRCh38
rs139132542 Chr.1: 78094805-78094805 on Build GRCh38
rs781542042 Chr.1: 78094968-78094968 on Build GRCh38
rs770530068 Chr.1: 78094951-78094951 on Build GRCh38
rs1012755027 Chr.1: 78094844-78094844 on Build GRCh38
rs565164 Chr.1: 78094899-78094899 on Build GRCh38
rs527759555 Chr.1: 78094822-78094822 on Build GRCh38
rs182786643 Chr.1: 78094937-78094937 on Build GRCh38

View Full Product Details