Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45472659-45472898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC1
Entrez Gene ID
2067
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
Gene Aliases
COFS4, RAD10, UV20
UniGene
Hs.435981
Gene Chromosome Location
Chr.19: 45407333-45478866 on Build GRCh38
Transcripts
XM_005258636.4 XM_017026464.1 NM_001114171.1 NM_006732.2 XM_005258691.1

Gene Symbol
FOSB
Entrez Gene ID
2354
Gene Name
FosB proto-oncogene, AP-1 transcription factor subunit
Gene Aliases
AP-1, G0S3, GOS3, GOSB
UniGene
Hs.590958
Gene Chromosome Location
Chr.19: 45467995-45475179 on Build GRCh38
Transcripts
XM_005258636.4 XM_017026464.1 NM_001114171.1 NM_006732.2 XM_005258691.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4846433 c.859C>G p.Q287E Chr.19: 45472854-45472854 on Build GRCh38
6666022 c.777G>A p.P259P Chr.19: 45472772-45472772 on Build GRCh38
6666020 c.896C>T p.S299L Chr.19: 45472891-45472891 on Build GRCh38
4685284 c.695C>T p.P232L Chr.19: 45472690-45472690 on Build GRCh38
998220 c.766C>T p.L256L Chr.19: 45472761-45472761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775259642 Chr.19: 45472687-45472687 on Build GRCh38
rs942024311 Chr.19: 45472679-45472679 on Build GRCh38
rs1051008029 Chr.19: 45472778-45472778 on Build GRCh38
rs781772652 Chr.19: 45472739-45472739 on Build GRCh38
rs765969336 Chr.19: 45472826-45472826 on Build GRCh38
rs868431295 Chr.19: 45472870-45472870 on Build GRCh38
rs896410074 Chr.19: 45472742-45472742 on Build GRCh38
rs1010929595 Chr.19: 45472772-45472772 on Build GRCh38
rs768599286 Chr.19: 45472696-45472696 on Build GRCh38
rs141121721 Chr.19: 45472805-45472805 on Build GRCh38
rs762720075 Chr.19: 45472817-45472817 on Build GRCh38
rs370863132 Chr.19: 45472721-45472721 on Build GRCh38
rs545320315 Chr.19: 45472843-45472843 on Build GRCh38
rs762369398 Chr.19: 45472729-45472729 on Build GRCh38
rs139625208 Chr.19: 45472864-45472864 on Build GRCh38
rs767156252 Chr.19: 45472815-45472815 on Build GRCh38
rs756602173 Chr.19: 45472756-45472756 on Build GRCh38
rs780808591 Chr.19: 45472764-45472778 on Build GRCh38
rs201294335 Chr.19: 45472868-45472868 on Build GRCh38
rs751040909 Chr.19: 45472835-45472835 on Build GRCh38
rs753320428 Chr.19: 45472863-45472863 on Build GRCh38
rs199730181 Chr.19: 45472704-45472704 on Build GRCh38
rs768081935 Chr.19: 45472859-45472859 on Build GRCh38
rs576246538 Chr.19: 45472733-45472733 on Build GRCh38
rs745962748 Chr.19: 45472800-45472800 on Build GRCh38
rs761585649 Chr.19: 45472812-45472812 on Build GRCh38
rs772711901 Chr.19: 45472816-45472816 on Build GRCh38
rs760716088 Chr.19: 45472693-45472693 on Build GRCh38
rs998592692 Chr.19: 45472775-45472775 on Build GRCh38
rs375283217 Chr.19: 45472822-45472822 on Build GRCh38
rs199545211 Chr.19: 45472825-45472825 on Build GRCh38
rs778997217 Chr.19: 45472783-45472783 on Build GRCh38
rs151110067 Chr.19: 45472754-45472754 on Build GRCh38
rs754172567 Chr.19: 45472871-45472871 on Build GRCh38
rs761597992 Chr.19: 45472720-45472720 on Build GRCh38
rs770893486 Chr.19: 45472780-45472780 on Build GRCh38
rs778135128 Chr.19: 45472758-45472758 on Build GRCh38
rs770758336 Chr.19: 45472682-45472682 on Build GRCh38
rs749563560 Chr.19: 45472767-45472767 on Build GRCh38
rs776560560 Chr.19: 45472809-45472809 on Build GRCh38

View Full Product Details