Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235342301-235342568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGPS1
Entrez Gene ID
9453
Gene Name
geranylgeranyl diphosphate synthase 1
Gene Aliases
GGPPS, GGPPS1
UniGene
Hs.730768
Gene Chromosome Location
Chr.1: 235328438-235344532 on Build GRCh38
Transcripts
NM_001037277.1 NR_036605.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6585634 c.683C>G p.T228S Chr.1: 235342552-235342552 on Build GRCh38
905828 c.525G>A p.P175P Chr.1: 235342394-235342394 on Build GRCh38
1748237 c.528A>G p.L176L Chr.1: 235342397-235342397 on Build GRCh38
425635 c.642A>G p.S214S Chr.1: 235342511-235342511 on Build GRCh38
4931074 c.470G>A p.G157E Chr.1: 235342339-235342339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772795288 Chr.1: 235342334-235342334 on Build GRCh38
rs553468287 Chr.1: 235342424-235342424 on Build GRCh38
rs746434869 Chr.1: 235342474-235342474 on Build GRCh38
rs371055539 Chr.1: 235342353-235342353 on Build GRCh38
rs146421372 Chr.1: 235342534-235342534 on Build GRCh38
rs760202011 Chr.1: 235342521-235342521 on Build GRCh38
rs764828996 Chr.1: 235342518-235342518 on Build GRCh38
rs367703615 Chr.1: 235342411-235342411 on Build GRCh38
rs749361864 Chr.1: 235342452-235342452 on Build GRCh38
rs761522290 Chr.1: 235342386-235342386 on Build GRCh38
rs765969703 Chr.1: 235342542-235342542 on Build GRCh38
rs770150533 Chr.1: 235342429-235342429 on Build GRCh38
rs749914612 Chr.1: 235342394-235342394 on Build GRCh38
rs202228389 Chr.1: 235342465-235342465 on Build GRCh38
rs373685421 Chr.1: 235342415-235342415 on Build GRCh38
rs772423462 Chr.1: 235342484-235342484 on Build GRCh38
rs779823837 Chr.1: 235342397-235342397 on Build GRCh38
rs772957103 Chr.1: 235342531-235342531 on Build GRCh38
rs775837661 Chr.1: 235342501-235342501 on Build GRCh38
rs374597891 Chr.1: 235342364-235342364 on Build GRCh38
rs771086310 Chr.1: 235342462-235342462 on Build GRCh38
rs910830334 Chr.1: 235342448-235342448 on Build GRCh38
rs753478069 Chr.1: 235342382-235342382 on Build GRCh38
rs751563825 Chr.1: 235342405-235342405 on Build GRCh38
rs988281633 Chr.1: 235342355-235342355 on Build GRCh38
rs761397259 Chr.1: 235342504-235342504 on Build GRCh38
rs764662240 Chr.1: 235342393-235342393 on Build GRCh38
rs777865527 Chr.1: 235342441-235342441 on Build GRCh38
rs758315333 Chr.1: 235342395-235342395 on Build GRCh38
rs868257623 Chr.1: 235342392-235342392 on Build GRCh38
rs766616817 Chr.1: 235342378-235342380 on Build GRCh38
rs774596015 Chr.1: 235342496-235342497 on Build GRCh38

View Full Product Details