Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 171870651-171870924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW11
Entrez Gene ID
23291
Gene Name
F-box and WD repeat domain containing 11
Gene Aliases
BTRC2, BTRCP2, FBW1B, FBXW1B, Fbw11, Hos
UniGene
Hs.484138
Gene Chromosome Location
Chr.5: 171861549-172006873 on Build GRCh38
Transcripts
NM_012300.2 NM_033644.2 NM_033645.2 XM_005265855.4 XM_005265856.2 XM_005265857.2 XM_005265858.2 XM_017009279.1 XM_017009280.1 XM_017009281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5903540 c.1239-8C>T p.? Chr.5: 171870866-171870866 on Build GRCh38
5903539 c.1278-8C>T p.? Chr.5: 171870866-171870866 on Build GRCh38
1065802 c.1340C>T p.A447V Chr.5: 171870757-171870757 on Build GRCh38
1696751 c.1301G>T p.R434L Chr.5: 171870796-171870796 on Build GRCh38
6662434 c.1336G>A p.V446I Chr.5: 171870800-171870800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371843289 Chr.5: 171870722-171870722 on Build GRCh38
rs755648448 Chr.5: 171870711-171870711 on Build GRCh38
rs746019925 Chr.5: 171870756-171870756 on Build GRCh38
rs900075588 Chr.5: 171870867-171870867 on Build GRCh38
rs748686392 Chr.5: 171870895-171870895 on Build GRCh38
rs1051318368 Chr.5: 171870786-171870786 on Build GRCh38
rs929013511 Chr.5: 171870711-171870711 on Build GRCh38
rs550104279 Chr.5: 171870716-171870716 on Build GRCh38
rs530269912 Chr.5: 171870714-171870714 on Build GRCh38
rs376089647 Chr.5: 171870765-171870765 on Build GRCh38
rs185771463 Chr.5: 171870712-171870712 on Build GRCh38
rs982884330 Chr.5: 171870686-171870686 on Build GRCh38
rs386694942 Chr.5: 171870712-171870714 on Build GRCh38
rs372414101 Chr.5: 171870846-171870846 on Build GRCh38
rs376094870 Chr.5: 171870829-171870829 on Build GRCh38
rs747089093 Chr.5: 171870819-171870819 on Build GRCh38
rs755865786 Chr.5: 171870723-171870723 on Build GRCh38
rs777554209 Chr.5: 171870731-171870731 on Build GRCh38
rs770226444 Chr.5: 171870897-171870897 on Build GRCh38
rs772272918 Chr.5: 171870779-171870779 on Build GRCh38
rs780195723 Chr.5: 171870784-171870784 on Build GRCh38
rs778754927 Chr.5: 171870873-171870873 on Build GRCh38
rs367632533 Chr.5: 171870710-171870710 on Build GRCh38
rs149858272 Chr.5: 171870843-171870843 on Build GRCh38

View Full Product Details