Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 133220669-133220942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTB-49A3.2
Entrez Gene ID
105379181
Gene Name
uncharacterized CTB-49A3.2
Gene Aliases
-
UniGene
Hs.586216
Gene Chromosome Location
Chr.5: 133209993-133226035 on Build GRCh38
Transcripts
NM_015082.1 XM_011543283.1 XM_011543284.1 XM_011543286.2 XM_011543287.2 XM_017009251.1

Gene Symbol
FSTL4
Entrez Gene ID
23105
Gene Name
follistatin like 4
Gene Aliases
-
UniGene
Hs.483390
Gene Chromosome Location
Chr.5: 133194827-133842066 on Build GRCh38
Transcripts
NM_015082.1 XM_011543283.1 XM_011543284.1 XM_011543286.2 XM_011543287.2 XM_017009251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4465543 c.1389C>T p.I463I Chr.5: 133220817-133220817 on Build GRCh38
1433117 c.1390G>A p.V464I Chr.5: 133220816-133220816 on Build GRCh38
6846357 c.1405G>A p.D469N Chr.5: 133220801-133220801 on Build GRCh38
6319569 c.1341C>A p.G447G Chr.5: 133220865-133220865 on Build GRCh38
7301875 c.1377C>T p.D459D Chr.5: 133220829-133220829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201737601 Chr.5: 133220828-133220828 on Build GRCh38
rs746418577 Chr.5: 133220818-133220818 on Build GRCh38
rs141735817 Chr.5: 133220826-133220826 on Build GRCh38
rs768709553 Chr.5: 133220760-133220760 on Build GRCh38
rs927386984 Chr.5: 133220876-133220876 on Build GRCh38
rs147996597 Chr.5: 133220816-133220816 on Build GRCh38
rs756627893 Chr.5: 133220882-133220882 on Build GRCh38
rs781030076 Chr.5: 133220914-133220914 on Build GRCh38
rs764822925 Chr.5: 133220848-133220848 on Build GRCh38
rs200675196 Chr.5: 133220701-133220701 on Build GRCh38
rs776578436 Chr.5: 133220841-133220841 on Build GRCh38
rs953273634 Chr.5: 133220819-133220819 on Build GRCh38
rs148816407 Chr.5: 133220754-133220754 on Build GRCh38
rs770196347 Chr.5: 133220755-133220755 on Build GRCh38
rs778629401 Chr.5: 133220717-133220717 on Build GRCh38
rs763731230 Chr.5: 133220859-133220859 on Build GRCh38
rs56304413 Chr.5: 133220740-133220740 on Build GRCh38
rs754502527 Chr.5: 133220708-133220708 on Build GRCh38
rs780331891 Chr.5: 133220886-133220886 on Build GRCh38
rs143505855 Chr.5: 133220805-133220805 on Build GRCh38
rs750917171 Chr.5: 133220871-133220871 on Build GRCh38
rs766582884 Chr.5: 133220858-133220858 on Build GRCh38
rs760523813 Chr.5: 133220799-133220799 on Build GRCh38
rs774687175 Chr.5: 133220775-133220775 on Build GRCh38
rs140221506 Chr.5: 133220831-133220831 on Build GRCh38
rs756468381 Chr.5: 133220825-133220825 on Build GRCh38
rs138517121 Chr.5: 133220749-133220749 on Build GRCh38
rs893149572 Chr.5: 133220770-133220770 on Build GRCh38
rs374882836 Chr.5: 133220812-133220812 on Build GRCh38
rs767927742 Chr.5: 133220829-133220829 on Build GRCh38
rs778053204 Chr.5: 133220916-133220916 on Build GRCh38
rs754733229 Chr.5: 133220809-133220809 on Build GRCh38
rs748441796 Chr.5: 133220911-133220911 on Build GRCh38
rs937391930 Chr.5: 133220887-133220887 on Build GRCh38
rs772991203 Chr.5: 133220833-133220833 on Build GRCh38
rs756277645 Chr.5: 133220735-133220736 on Build GRCh38
rs552403197 Chr.5: 133220810-133220810 on Build GRCh38
rs370030673 Chr.5: 133220907-133220907 on Build GRCh38
rs989214230 Chr.5: 133220817-133220817 on Build GRCh38
rs760757129 Chr.5: 133220834-133220834 on Build GRCh38
rs370187282 Chr.5: 133220783-133220783 on Build GRCh38
rs1054412623 Chr.5: 133220767-133220767 on Build GRCh38
rs759189638 Chr.5: 133220844-133220844 on Build GRCh38
rs771351696 Chr.5: 133220727-133220727 on Build GRCh38
rs747534469 Chr.5: 133220723-133220723 on Build GRCh38
rs766313602 Chr.5: 133220800-133220800 on Build GRCh38
rs553294954 Chr.5: 133220908-133220908 on Build GRCh38
rs753592417 Chr.5: 133220705-133220705 on Build GRCh38
rs560378423 Chr.5: 133220695-133220695 on Build GRCh38
rs201153166 Chr.5: 133220835-133220835 on Build GRCh38
rs774310162 Chr.5: 133220763-133220763 on Build GRCh38
rs375819704 Chr.5: 133220849-133220849 on Build GRCh38
rs755786720 Chr.5: 133220698-133220698 on Build GRCh38
rs775661811 Chr.5: 133220757-133220757 on Build GRCh38
rs199589963 Chr.5: 133220832-133220832 on Build GRCh38
rs772036412 Chr.5: 133220912-133220912 on Build GRCh38
rs375077200 Chr.5: 133220759-133220759 on Build GRCh38
rs377420733 Chr.5: 133220769-133220769 on Build GRCh38

View Full Product Details