Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56472581-56472854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLS2
Entrez Gene ID
27165
Gene Name
glutaminase 2
Gene Aliases
GA, GLS, LGA, hLGA
UniGene
Hs.212606
Gene Chromosome Location
Chr.12: 56470944-56488414 on Build GRCh38
Transcripts
NM_001280796.1 NM_001280797.1 NM_001280798.1 NM_013267.3 XM_005268797.1 XM_017019180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1628726 c.1454A>C p.K485T Chr.12: 56472747-56472747 on Build GRCh38
941700 c.1491C>T p.G497G Chr.12: 56472710-56472710 on Build GRCh38
3376441 c.1484A>G p.Y495C Chr.12: 56472717-56472717 on Build GRCh38
6031657 c.1508G>A p.R503Q Chr.12: 56472693-56472693 on Build GRCh38
4188691 c.1501G>T p.A501S Chr.12: 56472700-56472700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369790479 Chr.12: 56472690-56472690 on Build GRCh38
rs982516597 Chr.12: 56472709-56472709 on Build GRCh38
rs772287526 Chr.12: 56472789-56472789 on Build GRCh38
rs751104669 Chr.12: 56472739-56472739 on Build GRCh38
rs111276307 Chr.12: 56472738-56472738 on Build GRCh38
rs538737952 Chr.12: 56472607-56472607 on Build GRCh38
rs769116572 Chr.12: 56472772-56472772 on Build GRCh38
rs767761248 Chr.12: 56472654-56472654 on Build GRCh38
rs773489074 Chr.12: 56472790-56472790 on Build GRCh38
rs374417630 Chr.12: 56472765-56472765 on Build GRCh38
rs778777549 Chr.12: 56472672-56472676 on Build GRCh38
rs760752446 Chr.12: 56472802-56472802 on Build GRCh38
rs377467340 Chr.12: 56472771-56472771 on Build GRCh38
rs936011017 Chr.12: 56472641-56472641 on Build GRCh38
rs745844095 Chr.12: 56472678-56472679 on Build GRCh38
rs536675044 Chr.12: 56472671-56472671 on Build GRCh38
rs760843313 Chr.12: 56472683-56472683 on Build GRCh38
rs779303058 Chr.12: 56472774-56472774 on Build GRCh38
rs542322164 Chr.12: 56472623-56472623 on Build GRCh38
rs754953103 Chr.12: 56472694-56472694 on Build GRCh38
rs773260635 Chr.12: 56472667-56472667 on Build GRCh38
rs748572561 Chr.12: 56472776-56472776 on Build GRCh38
rs906384806 Chr.12: 56472821-56472821 on Build GRCh38
rs756801839 Chr.12: 56472762-56472762 on Build GRCh38
rs771193153 Chr.12: 56472803-56472803 on Build GRCh38
rs572058296 Chr.12: 56472620-56472620 on Build GRCh38
rs753842932 Chr.12: 56472693-56472693 on Build GRCh38
rs867382834 Chr.12: 56472822-56472822 on Build GRCh38
rs761985241 Chr.12: 56472650-56472650 on Build GRCh38
rs3211277 Chr.12: 56472697-56472697 on Build GRCh38
rs761742311 Chr.12: 56472685-56472685 on Build GRCh38
rs560712128 Chr.12: 56472717-56472717 on Build GRCh38
rs139745938 Chr.12: 56472734-56472734 on Build GRCh38
rs752514413 Chr.12: 56472707-56472707 on Build GRCh38
rs765192271 Chr.12: 56472705-56472705 on Build GRCh38

View Full Product Details