Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76075091-76075362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALR2
Entrez Gene ID
8811
Gene Name
galanin receptor 2
Gene Aliases
GAL2-R, GALNR2, GALR-2
UniGene
Hs.666366
Gene Chromosome Location
Chr.17: 76070822-76077541 on Build GRCh38
Transcripts
NM_003857.3 XM_011525427.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4069792 c.334A>G p.S112G Chr.17: 76075217-76075217 on Build GRCh38
6669181 c.339C>T p.S113S Chr.17: 76075222-76075222 on Build GRCh38
7369187 c.247G>A p.A83T Chr.17: 76075130-76075130 on Build GRCh38
3388150 c.247G>T p.A83S Chr.17: 76075130-76075130 on Build GRCh38
2742878 c.310C>T p.L104F Chr.17: 76075193-76075193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372156922 Chr.17: 76075285-76075285 on Build GRCh38
rs886236002 Chr.17: 76075256-76075256 on Build GRCh38
rs762099186 Chr.17: 76075124-76075124 on Build GRCh38
rs1053234576 Chr.17: 76075201-76075201 on Build GRCh38
rs759023292 Chr.17: 76075270-76075270 on Build GRCh38
rs893216157 Chr.17: 76075206-76075206 on Build GRCh38
rs139644215 Chr.17: 76075140-76075140 on Build GRCh38
rs948969035 Chr.17: 76075135-76075135 on Build GRCh38
rs752548430 Chr.17: 76075214-76075214 on Build GRCh38
rs745640817 Chr.17: 76075272-76075272 on Build GRCh38
rs990525594 Chr.17: 76075296-76075296 on Build GRCh38
rs751058967 Chr.17: 76075175-76075175 on Build GRCh38
rs758621186 Chr.17: 76075233-76075233 on Build GRCh38
rs755918092 Chr.17: 76075227-76075227 on Build GRCh38
rs534498888 Chr.17: 76075234-76075234 on Build GRCh38
rs772801215 Chr.17: 76075136-76075136 on Build GRCh38
rs1040766068 Chr.17: 76075185-76075185 on Build GRCh38
rs982061121 Chr.17: 76075313-76075313 on Build GRCh38
rs150837200 Chr.17: 76075228-76075228 on Build GRCh38
rs765848984 Chr.17: 76075173-76075173 on Build GRCh38
rs750648346 Chr.17: 76075231-76075231 on Build GRCh38
rs143605647 Chr.17: 76075130-76075130 on Build GRCh38
rs578111238 Chr.17: 76075295-76075295 on Build GRCh38
rs1020242539 Chr.17: 76075321-76075321 on Build GRCh38
rs375568757 Chr.17: 76075268-76075268 on Build GRCh38
rs748534104 Chr.17: 76075248-76075248 on Build GRCh38
rs368215026 Chr.17: 76075269-76075269 on Build GRCh38
rs116466433 Chr.17: 76075332-76075332 on Build GRCh38
rs967326285 Chr.17: 76075330-76075330 on Build GRCh38
rs997812315 Chr.17: 76075236-76075236 on Build GRCh38
rs199942975 Chr.17: 76075299-76075299 on Build GRCh38
rs141784016 Chr.17: 76075213-76075213 on Build GRCh38
rs769155155 Chr.17: 76075237-76075237 on Build GRCh38
rs993937166 Chr.17: 76075221-76075221 on Build GRCh38
rs781620715 Chr.17: 76075242-76075242 on Build GRCh38
rs375532301 Chr.17: 76075257-76075257 on Build GRCh38
rs370909338 Chr.17: 76075210-76075210 on Build GRCh38
rs762561971 Chr.17: 76075262-76075262 on Build GRCh38
rs770105998 Chr.17: 76075251-76075251 on Build GRCh38
rs970349635 Chr.17: 76075306-76075306 on Build GRCh38
rs990885357 Chr.17: 76075122-76075122 on Build GRCh38

View Full Product Details