Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154584274-154584505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGA
Entrez Gene ID
2243
Gene Name
fibrinogen alpha chain
Gene Aliases
Fib2
UniGene
Hs.351593
Gene Chromosome Location
Chr.4: 154583126-154590766 on Build GRCh38
Transcripts
NM_000508.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
311164 c.2416G>A p.G806S Chr.4: 154584309-154584309 on Build GRCh38
4934054 c.2226C>G p.F742L Chr.4: 154584499-154584499 on Build GRCh38
5531226 c.2445C>A p.A815A Chr.4: 154584280-154584280 on Build GRCh38
4820252 c.2391G>C p.E797D Chr.4: 154584334-154584334 on Build GRCh38
1052409 c.2237C>A p.S746Y Chr.4: 154584488-154584488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780618617 Chr.4: 154584415-154584415 on Build GRCh38
rs955756291 Chr.4: 154584318-154584318 on Build GRCh38
rs765888222 Chr.4: 154584402-154584402 on Build GRCh38
rs768839775 Chr.4: 154584332-154584332 on Build GRCh38
rs769797158 Chr.4: 154584436-154584436 on Build GRCh38
rs763593060 Chr.4: 154584386-154584386 on Build GRCh38
rs769039996 Chr.4: 154584425-154584425 on Build GRCh38
rs899420855 Chr.4: 154584445-154584445 on Build GRCh38
rs771538227 Chr.4: 154584310-154584310 on Build GRCh38
rs190999281 Chr.4: 154584439-154584439 on Build GRCh38
rs777521200 Chr.4: 154584476-154584476 on Build GRCh38
rs755826305 Chr.4: 154584385-154584385 on Build GRCh38
rs747229356 Chr.4: 154584419-154584419 on Build GRCh38
rs756100934 Chr.4: 154584450-154584453 on Build GRCh38
rs556540003 Chr.4: 154584336-154584336 on Build GRCh38
rs775785750 Chr.4: 154584324-154584324 on Build GRCh38
rs762767715 Chr.4: 154584345-154584345 on Build GRCh38
rs376739537 Chr.4: 154584473-154584473 on Build GRCh38
rs753431992 Chr.4: 154584394-154584394 on Build GRCh38
rs200427267 Chr.4: 154584322-154584322 on Build GRCh38
rs747243710 Chr.4: 154584325-154584325 on Build GRCh38
rs980896093 Chr.4: 154584393-154584393 on Build GRCh38
rs374692754 Chr.4: 154584359-154584359 on Build GRCh38
rs150073296 Chr.4: 154584440-154584440 on Build GRCh38
rs371718702 Chr.4: 154584452-154584452 on Build GRCh38
rs752625164 Chr.4: 154584378-154584380 on Build GRCh38
rs762189767 Chr.4: 154584380-154584380 on Build GRCh38
rs774179478 Chr.4: 154584367-154584367 on Build GRCh38
rs368008640 Chr.4: 154584481-154584481 on Build GRCh38
rs748208594 Chr.4: 154584435-154584435 on Build GRCh38
rs918210100 Chr.4: 154584337-154584337 on Build GRCh38
rs776549090 Chr.4: 154584427-154584427 on Build GRCh38
rs756717254 Chr.4: 154584455-154584455 on Build GRCh38
rs779119250 Chr.4: 154584315-154584315 on Build GRCh38
rs367614797 Chr.4: 154584414-154584414 on Build GRCh38
rs749749951 Chr.4: 154584308-154584308 on Build GRCh38
rs753458290 Chr.4: 154584453-154584453 on Build GRCh38
rs769643431 Chr.4: 154584429-154584429 on Build GRCh38
rs148824832 Chr.4: 154584375-154584375 on Build GRCh38
rs764664187 Chr.4: 154584471-154584471 on Build GRCh38
rs774280341 Chr.4: 154584437-154584437 on Build GRCh38
rs971302509 Chr.4: 154584377-154584377 on Build GRCh38
rs775334799 Chr.4: 154584444-154584444 on Build GRCh38
rs777278632 Chr.4: 154584305-154584305 on Build GRCh38
rs758042722 Chr.4: 154584463-154584463 on Build GRCh38
rs779432672 Chr.4: 154584409-154584409 on Build GRCh38
rs569852674 Chr.4: 154584350-154584350 on Build GRCh38
rs767304497 Chr.4: 154584376-154584376 on Build GRCh38

View Full Product Details