Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16256549-16256816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO42
Entrez Gene ID
54455
Gene Name
F-box protein 42
Gene Aliases
Fbx42, JFK
UniGene
Hs.522384
Gene Chromosome Location
Chr.1: 16246840-16352491 on Build GRCh38
Transcripts
NM_018994.2 XM_006710698.3 XM_017001502.1 XM_017001503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4024749 c.583C>T p.P195S Chr.1: 16256679-16256679 on Build GRCh38
3477663 c.511C>T p.P171S Chr.1: 16256751-16256751 on Build GRCh38
6019403 c.632_643del12 p.H211_S214delHTYS Chr.1: 16256619-16256630 on Build GRCh38
3477662 c.512C>T p.P171L Chr.1: 16256750-16256750 on Build GRCh38
5585197 c.540C>T p.V180V Chr.1: 16256722-16256722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745717475 Chr.1: 16256745-16256745 on Build GRCh38
rs775269054 Chr.1: 16256623-16256623 on Build GRCh38
rs758421477 Chr.1: 16256727-16256727 on Build GRCh38
rs763594734 Chr.1: 16256682-16256682 on Build GRCh38
rs1016115246 Chr.1: 16256717-16256717 on Build GRCh38
rs768076976 Chr.1: 16256788-16256788 on Build GRCh38
rs368638977 Chr.1: 16256683-16256683 on Build GRCh38
rs769711077 Chr.1: 16256746-16256746 on Build GRCh38
rs533604479 Chr.1: 16256731-16256731 on Build GRCh38
rs34339996 Chr.1: 16256663-16256664 on Build GRCh38
rs761199756 Chr.1: 16256695-16256695 on Build GRCh38
rs370415316 Chr.1: 16256662-16256662 on Build GRCh38
rs761450488 Chr.1: 16256619-16256622 on Build GRCh38
rs372330593 Chr.1: 16256721-16256721 on Build GRCh38
rs375012254 Chr.1: 16256722-16256722 on Build GRCh38
rs775289100 Chr.1: 16256779-16256779 on Build GRCh38
rs966076502 Chr.1: 16256770-16256770 on Build GRCh38
rs746952518 Chr.1: 16256743-16256743 on Build GRCh38
rs1007654316 Chr.1: 16256724-16256724 on Build GRCh38
rs749964611 Chr.1: 16256790-16256791 on Build GRCh38
rs148204328 Chr.1: 16256585-16256585 on Build GRCh38
rs769621255 Chr.1: 16256602-16256602 on Build GRCh38
rs202166367 Chr.1: 16256725-16256725 on Build GRCh38
rs766761082 Chr.1: 16256704-16256704 on Build GRCh38
rs759374568 Chr.1: 16256587-16256587 on Build GRCh38
rs748895242 Chr.1: 16256781-16256781 on Build GRCh38
rs767011648 Chr.1: 16256774-16256775 on Build GRCh38
rs758766797 Chr.1: 16256667-16256667 on Build GRCh38
rs770652643 Chr.1: 16256581-16256581 on Build GRCh38
rs376029455 Chr.1: 16256681-16256681 on Build GRCh38
rs972473378 Chr.1: 16256782-16256782 on Build GRCh38

View Full Product Details