Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15272242-15272471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHAD1
Entrez Gene ID
114827
Gene Name
forkhead associated phosphopeptide binding domain 1
Gene Aliases
-
UniGene
Hs.659997
Gene Chromosome Location
Chr.1: 15236559-15400283 on Build GRCh38
Transcripts
NM_052929.1 XM_011540575.2 XM_011540576.2 XM_011540577.2 XM_011540580.2 XM_011540581.2 XM_011540582.2 XM_011540584.2 XM_011540588.2 XM_011540589.2 XM_011540590.2 XM_011540592.2 XM_011540593.2 XM_011540597.2 XM_011540598.2 XM_017000198.1 XM_017000199.1 XM_017000200.1 XM_017000201.1 XM_017000202.1 XM_017000203.1 XM_017000204.1 XM_017000205.1 XM_017000206.1 XM_017000207.1 XM_017000208.1 XM_017000209.1 XM_017000210.1 XM_017000211.1 XM_017000212.1 XM_017000213.1 XM_017000214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4222444 c.105C>T p.I35I Chr.1: 15272334-15272334 on Build GRCh38
6377716 c.157C>A p.L53I Chr.1: 15272386-15272386 on Build GRCh38
4951865 c.229A>G p.I77V Chr.1: 15272458-15272458 on Build GRCh38
4647355 c.195C>T p.N65N Chr.1: 15272424-15272424 on Build GRCh38
897096 c.118G>A p.A40T Chr.1: 15272347-15272347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369555756 Chr.1: 15272364-15272364 on Build GRCh38
rs959692341 Chr.1: 15272380-15272380 on Build GRCh38
rs756994252 Chr.1: 15272274-15272274 on Build GRCh38
rs534187667 Chr.1: 15272298-15272298 on Build GRCh38
rs990832284 Chr.1: 15272288-15272288 on Build GRCh38
rs765416342 Chr.1: 15272443-15272443 on Build GRCh38
rs890358230 Chr.1: 15272425-15272425 on Build GRCh38
rs902720384 Chr.1: 15272332-15272332 on Build GRCh38
rs139010451 Chr.1: 15272265-15272265 on Build GRCh38
rs991176014 Chr.1: 15272389-15272389 on Build GRCh38
rs915800574 Chr.1: 15272415-15272415 on Build GRCh38
rs114094133 Chr.1: 15272299-15272299 on Build GRCh38
rs771122194 Chr.1: 15272390-15272390 on Build GRCh38
rs894088523 Chr.1: 15272315-15272315 on Build GRCh38
rs908372567 Chr.1: 15272444-15272444 on Build GRCh38
rs12061399 Chr.1: 15272316-15272316 on Build GRCh38
rs776954896 Chr.1: 15272399-15272399 on Build GRCh38
rs182131431 Chr.1: 15272335-15272335 on Build GRCh38
rs1015166889 Chr.1: 15272434-15272434 on Build GRCh38
rs368274850 Chr.1: 15272286-15272286 on Build GRCh38
rs747187763 Chr.1: 15272369-15272369 on Build GRCh38
rs931267727 Chr.1: 15272277-15272277 on Build GRCh38
rs779278948 Chr.1: 15272306-15272306 on Build GRCh38
rs920010853 Chr.1: 15272273-15272273 on Build GRCh38
rs772727638 Chr.1: 15272370-15272370 on Build GRCh38
rs571963196 Chr.1: 15272270-15272270 on Build GRCh38
rs996835210 Chr.1: 15272304-15272306 on Build GRCh38
rs995776529 Chr.1: 15272374-15272374 on Build GRCh38
rs1008847365 Chr.1: 15272431-15272431 on Build GRCh38
rs773593901 Chr.1: 15272358-15272358 on Build GRCh38
rs769287042 Chr.1: 15272334-15272334 on Build GRCh38
rs556223963 Chr.1: 15272424-15272424 on Build GRCh38
rs967494302 Chr.1: 15272357-15272357 on Build GRCh38

View Full Product Details