Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 176221148-176221409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB5
Entrez Gene ID
140458
Gene Name
ankyrin repeat and SOCS box containing 5
Gene Aliases
ASB-5
UniGene
Hs.591712
Gene Chromosome Location
Chr.4: 176213673-176277571 on Build GRCh38
Transcripts
NM_080874.3 XM_005262759.1 XM_011531616.2 XM_011531617.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4135501 c.627T>A p.C209* Chr.4: 176221198-176221198 on Build GRCh38
6166971 c.581T>A p.V194D Chr.4: 176221244-176221244 on Build GRCh38
6166972 c.611C>T p.P204L Chr.4: 176221214-176221214 on Build GRCh38
1053426 c.661C>A p.L221I Chr.4: 176221164-176221164 on Build GRCh38
4774403 c.575T>C p.I192T Chr.4: 176221250-176221250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867841129 Chr.4: 176221256-176221256 on Build GRCh38
rs766191366 Chr.4: 176221258-176221258 on Build GRCh38
rs902594228 Chr.4: 176221225-176221225 on Build GRCh38
rs145043214 Chr.4: 176221288-176221288 on Build GRCh38
rs1052214117 Chr.4: 176221247-176221247 on Build GRCh38
rs761591211 Chr.4: 176221214-176221214 on Build GRCh38
rs1003817949 Chr.4: 176221245-176221245 on Build GRCh38
rs906434029 Chr.4: 176221239-176221239 on Build GRCh38
rs764178247 Chr.4: 176221281-176221281 on Build GRCh38
rs570156844 Chr.4: 176221333-176221333 on Build GRCh38
rs371930383 Chr.4: 176221292-176221292 on Build GRCh38
rs796087305 Chr.4: 176221226-176221226 on Build GRCh38
rs1030113478 Chr.4: 176221348-176221348 on Build GRCh38
rs765314123 Chr.4: 176221299-176221299 on Build GRCh38
rs755503780 Chr.4: 176221326-176221326 on Build GRCh38
rs762714429 Chr.4: 176221255-176221255 on Build GRCh38
rs772805804 Chr.4: 176221336-176221336 on Build GRCh38
rs141251672 Chr.4: 176221270-176221270 on Build GRCh38
rs751490732 Chr.4: 176221260-176221260 on Build GRCh38
rs963509607 Chr.4: 176221376-176221376 on Build GRCh38
rs947714524 Chr.4: 176221209-176221209 on Build GRCh38
rs950477105 Chr.4: 176221358-176221358 on Build GRCh38
rs780447709 Chr.4: 176221324-176221324 on Build GRCh38
rs954975397 Chr.4: 176221368-176221368 on Build GRCh38
rs746147996 Chr.4: 176221175-176221175 on Build GRCh38
rs772475742 Chr.4: 176221183-176221183 on Build GRCh38
rs990699062 Chr.4: 176221381-176221381 on Build GRCh38
rs116013117 Chr.4: 176221374-176221374 on Build GRCh38
rs376606057 Chr.4: 176221321-176221321 on Build GRCh38
rs1021630943 Chr.4: 176221323-176221323 on Build GRCh38
rs142765630 Chr.4: 176221201-176221201 on Build GRCh38
rs151040450 Chr.4: 176221212-176221212 on Build GRCh38
rs12501772 Chr.4: 176221375-176221375 on Build GRCh38
rs138746920 Chr.4: 176221240-176221240 on Build GRCh38
rs34466424 Chr.4: 176221337-176221337 on Build GRCh38
rs770678956 Chr.4: 176221339-176221339 on Build GRCh38
rs773073544 Chr.4: 176221241-176221241 on Build GRCh38
rs768333542 Chr.4: 176221203-176221203 on Build GRCh38
rs141181389 Chr.4: 176221341-176221341 on Build GRCh38
rs570133744 Chr.4: 176221325-176221325 on Build GRCh38
rs750657946 Chr.4: 176221308-176221308 on Build GRCh38
rs780855507 Chr.4: 176221332-176221332 on Build GRCh38

View Full Product Details