Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132342517-132342790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACPP
Entrez Gene ID
55
Gene Name
acid phosphatase, prostate
Gene Aliases
5'-NT, ACP-3, ACP3
UniGene
Hs.433060
Gene Chromosome Location
Chr.3: 132317367-132368765 on Build GRCh38
Transcripts
NM_001099.4 NM_001134194.1 NM_001292037.1 XM_011512946.1 XM_011512947.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919611 c.556-2A>C p.? Chr.3: 132342550-132342550 on Build GRCh38
1038384 c.556-1G>T p.? Chr.3: 132342551-132342551 on Build GRCh38
3587557 c.642T>G p.Y214* Chr.3: 132342638-132342638 on Build GRCh38
1038386 c.631G>A p.D211N Chr.3: 132342627-132342627 on Build GRCh38
1616918 c.648+7_648+8insA p.? Chr.3: 132342651-132342652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996871169 Chr.3: 132342709-132342709 on Build GRCh38
rs751367677 Chr.3: 132342614-132342626 on Build GRCh38
rs959665839 Chr.3: 132342564-132342564 on Build GRCh38
rs760770327 Chr.3: 132342590-132342590 on Build GRCh38
rs776760430 Chr.3: 132342682-132342682 on Build GRCh38
rs747168242 Chr.3: 132342672-132342672 on Build GRCh38
rs749813514 Chr.3: 132342639-132342639 on Build GRCh38
rs370686017 Chr.3: 132342692-132342692 on Build GRCh38
rs748915316 Chr.3: 132342739-132342739 on Build GRCh38
rs759459732 Chr.3: 132342688-132342688 on Build GRCh38
rs755502254 Chr.3: 132342642-132342642 on Build GRCh38
rs777995289 Chr.3: 132342652-132342652 on Build GRCh38
rs751215762 Chr.3: 132342626-132342626 on Build GRCh38
rs202202968 Chr.3: 132342651-132342652 on Build GRCh38
rs773332320 Chr.3: 132342588-132342588 on Build GRCh38
rs776438843 Chr.3: 132342605-132342605 on Build GRCh38
rs565585764 Chr.3: 132342738-132342738 on Build GRCh38
rs748438402 Chr.3: 132342578-132342578 on Build GRCh38
rs777524768 Chr.3: 132342627-132342627 on Build GRCh38
rs890320216 Chr.3: 132342599-132342599 on Build GRCh38
rs770930439 Chr.3: 132342674-132342674 on Build GRCh38
rs780853788 Chr.3: 132342570-132342570 on Build GRCh38
rs748493315 Chr.3: 132342661-132342661 on Build GRCh38
rs753937634 Chr.3: 132342681-132342682 on Build GRCh38
rs752507428 Chr.3: 132342612-132342612 on Build GRCh38
rs755850776 Chr.3: 132342614-132342614 on Build GRCh38
rs774762429 Chr.3: 132342549-132342549 on Build GRCh38
rs779187360 Chr.3: 132342656-132342656 on Build GRCh38
rs376742673 Chr.3: 132342683-132342684 on Build GRCh38
rs752477546 Chr.3: 132342600-132342600 on Build GRCh38
rs780691182 Chr.3: 132342630-132342630 on Build GRCh38
rs759461213 Chr.3: 132342608-132342608 on Build GRCh38
rs763859920 Chr.3: 132342616-132342616 on Build GRCh38
rs772254420 Chr.3: 132342579-132342579 on Build GRCh38
rs918920096 Chr.3: 132342562-132342562 on Build GRCh38
rs916142235 Chr.3: 132342679-132342679 on Build GRCh38
rs576994797 Chr.3: 132342751-132342751 on Build GRCh38
rs773458707 Chr.3: 132342666-132342666 on Build GRCh38

View Full Product Details