Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 20643127-20643358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB5
Entrez Gene ID
340273
Gene Name
ATP binding cassette subfamily B member 5
Gene Aliases
ABCB5alpha, ABCB5beta, EST422562
UniGene
Hs.404102
Gene Chromosome Location
Chr.7: 20615622-20757014 on Build GRCh38
Transcripts
NM_001163941.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3663082 c.415A>T p.I139F Chr.7: 20643284-20643284 on Build GRCh38
3923333 c.401G>A p.R134Q Chr.7: 20643270-20643270 on Build GRCh38
5062370 c.426_427insT p.H145fs*12 Chr.7: 20643295-20643296 on Build GRCh38
7353574 c.370C>G p.Q124E Chr.7: 20643239-20643239 on Build GRCh38
6563690 c.488T>A p.L163H Chr.7: 20643357-20643357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77409024 Chr.7: 20643332-20643332 on Build GRCh38
rs202100779 Chr.7: 20643234-20643234 on Build GRCh38
rs772896092 Chr.7: 20643273-20643273 on Build GRCh38
rs186728686 Chr.7: 20643327-20643327 on Build GRCh38
rs950647261 Chr.7: 20643216-20643216 on Build GRCh38
rs565994506 Chr.7: 20643279-20643279 on Build GRCh38
rs763832811 Chr.7: 20643160-20643160 on Build GRCh38
rs747180120 Chr.7: 20643217-20643217 on Build GRCh38
rs766371098 Chr.7: 20643282-20643282 on Build GRCh38
rs753647911 Chr.7: 20643287-20643287 on Build GRCh38
rs372749249 Chr.7: 20643238-20643238 on Build GRCh38
rs887634404 Chr.7: 20643211-20643211 on Build GRCh38
rs757514065 Chr.7: 20643302-20643302 on Build GRCh38
rs769194692 Chr.7: 20643331-20643331 on Build GRCh38
rs770339087 Chr.7: 20643251-20643251 on Build GRCh38
rs372132207 Chr.7: 20643315-20643315 on Build GRCh38
rs548933985 Chr.7: 20643325-20643325 on Build GRCh38
rs757454697 Chr.7: 20643245-20643245 on Build GRCh38
rs766397154 Chr.7: 20643184-20643184 on Build GRCh38
rs141511940 Chr.7: 20643269-20643269 on Build GRCh38
rs570793675 Chr.7: 20643324-20643324 on Build GRCh38
rs918665949 Chr.7: 20643159-20643159 on Build GRCh38
rs767080146 Chr.7: 20643172-20643172 on Build GRCh38
rs374303845 Chr.7: 20643277-20643277 on Build GRCh38
rs761412272 Chr.7: 20643169-20643169 on Build GRCh38
rs745974541 Chr.7: 20643323-20643323 on Build GRCh38
rs1047512159 Chr.7: 20643199-20643199 on Build GRCh38
rs548365815 Chr.7: 20643249-20643249 on Build GRCh38
rs199687406 Chr.7: 20643253-20643253 on Build GRCh38
rs946181751 Chr.7: 20643281-20643281 on Build GRCh38
rs377268116 Chr.7: 20643162-20643162 on Build GRCh38
rs181716362 Chr.7: 20643326-20643326 on Build GRCh38
rs756158660 Chr.7: 20643179-20643179 on Build GRCh38
rs190460902 Chr.7: 20643270-20643270 on Build GRCh38
rs368226947 Chr.7: 20643288-20643288 on Build GRCh38
rs375764523 Chr.7: 20643250-20643250 on Build GRCh38
rs369274708 Chr.7: 20643180-20643180 on Build GRCh38
rs754685763 Chr.7: 20643212-20643212 on Build GRCh38
rs774068423 Chr.7: 20643262-20643262 on Build GRCh38
rs1035362873 Chr.7: 20643252-20643252 on Build GRCh38
rs771871738 Chr.7: 20643335-20643335 on Build GRCh38
rs17143212 Chr.7: 20643261-20643261 on Build GRCh38

View Full Product Details