Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103083649-103083874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC10
Entrez Gene ID
83787
Gene Name
armadillo repeat containing 10
Gene Aliases
PNAS-112, PNAS112, PSEC0198, SVH
UniGene
Hs.287412
Gene Chromosome Location
Chr.7: 103074727-103099763 on Build GRCh38
Transcripts
NM_001161009.2 NM_001161010.2 NM_001161011.2 NM_001161012.2 NM_001161013.2 NM_031905.4 XM_005250628.2 XM_011516601.2 XM_011516602.2 XM_017012684.1 XM_017012685.1 XM_017012686.1 XM_017012687.1 XM_017012688.1 XM_017012689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3083677 c.258T>A p.D86E Chr.7: 103083695-103083695 on Build GRCh38
4625885 c.333T>C p.P111P Chr.7: 103083770-103083770 on Build GRCh38
6432292 c.389A>T p.N130I Chr.7: 103083826-103083826 on Build GRCh38
3995220 c.346A>G p.R116G Chr.7: 103083783-103083783 on Build GRCh38
5908720 c.263C>T p.S88L Chr.7: 103083700-103083700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056921270 Chr.7: 103083778-103083778 on Build GRCh38
rs780940368 Chr.7: 103083694-103083694 on Build GRCh38
rs1052782940 Chr.7: 103083723-103083723 on Build GRCh38
rs771574664 Chr.7: 103083712-103083713 on Build GRCh38
rs754872562 Chr.7: 103083796-103083796 on Build GRCh38
rs553957098 Chr.7: 103083732-103083732 on Build GRCh38
rs777375435 Chr.7: 103083815-103083815 on Build GRCh38
rs962986025 Chr.7: 103083804-103083804 on Build GRCh38
rs762906223 Chr.7: 103083775-103083775 on Build GRCh38
rs145780869 Chr.7: 103083830-103083830 on Build GRCh38
rs767066870 Chr.7: 103083811-103083811 on Build GRCh38
rs746382759 Chr.7: 103083719-103083719 on Build GRCh38
rs200030413 Chr.7: 103083794-103083794 on Build GRCh38
rs772621980 Chr.7: 103083731-103083732 on Build GRCh38
rs151325875 Chr.7: 103083686-103083686 on Build GRCh38
rs772526195 Chr.7: 103083722-103083722 on Build GRCh38
rs776977533 Chr.7: 103083759-103083759 on Build GRCh38
rs747610756 Chr.7: 103083706-103083706 on Build GRCh38
rs374014963 Chr.7: 103083761-103083761 on Build GRCh38
rs769407196 Chr.7: 103083708-103083708 on Build GRCh38
rs891426942 Chr.7: 103083740-103083740 on Build GRCh38
rs189688478 Chr.7: 103083739-103083739 on Build GRCh38
rs768727142 Chr.7: 103083751-103083751 on Build GRCh38
rs761785012 Chr.7: 103083760-103083760 on Build GRCh38
rs1017549708 Chr.7: 103083797-103083797 on Build GRCh38
rs766111577 Chr.7: 103083786-103083786 on Build GRCh38
rs752412153 Chr.7: 103083814-103083814 on Build GRCh38
rs28700061 Chr.7: 103083763-103083763 on Build GRCh38
rs773063738 Chr.7: 103083774-103083774 on Build GRCh38
rs141475643 Chr.7: 103083717-103083717 on Build GRCh38

View Full Product Details