Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 141292718-141292953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PXYLP1
Entrez Gene ID
92370
Gene Name
2-phosphoxylose phosphatase 1
Gene Aliases
ACPL2, HEL124, XYLP
UniGene
Hs.657887
Gene Chromosome Location
Chr.3: 141231825-141294903 on Build GRCh38
Transcripts
NM_001037172.2 NM_001282728.1 NM_152282.4 XM_005247903.3 XM_011513314.2 XM_017007510.1 XM_017007511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1752941 c.1158C>T p.L386L Chr.3: 141292920-141292920 on Build GRCh38
4114042 c.1085G>A p.R362H Chr.3: 141292847-141292847 on Build GRCh38
4114044 c.1125C>T p.Y375Y Chr.3: 141292887-141292887 on Build GRCh38
4851013 c.1045G>A p.E349K Chr.3: 141292855-141292855 on Build GRCh38
3846385 c.1044C>T p.T348T Chr.3: 141292854-141292854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894477517 Chr.3: 141292922-141292922 on Build GRCh38
rs760035675 Chr.3: 141292880-141292880 on Build GRCh38
rs755822898 Chr.3: 141292884-141292884 on Build GRCh38
rs747227880 Chr.3: 141292768-141292768 on Build GRCh38
rs115983278 Chr.3: 141292910-141292910 on Build GRCh38
rs756960394 Chr.3: 141292824-141292824 on Build GRCh38
rs775346386 Chr.3: 141292804-141292804 on Build GRCh38
rs142753948 Chr.3: 141292855-141292855 on Build GRCh38
rs750419352 Chr.3: 141292911-141292911 on Build GRCh38
rs775151612 Chr.3: 141292874-141292874 on Build GRCh38
rs770036809 Chr.3: 141292853-141292853 on Build GRCh38
rs147978864 Chr.3: 141292933-141292933 on Build GRCh38
rs533413899 Chr.3: 141292839-141292839 on Build GRCh38
rs758424483 Chr.3: 141292756-141292756 on Build GRCh38
rs114644481 Chr.3: 141292912-141292912 on Build GRCh38
rs763593805 Chr.3: 141292808-141292808 on Build GRCh38
rs188165694 Chr.3: 141292900-141292900 on Build GRCh38
rs771763179 Chr.3: 141292858-141292858 on Build GRCh38
rs541033754 Chr.3: 141292827-141292827 on Build GRCh38
rs773892481 Chr.3: 141292854-141292854 on Build GRCh38
rs754036846 Chr.3: 141292909-141292910 on Build GRCh38
rs764353806 Chr.3: 141292822-141292826 on Build GRCh38
rs115260575 Chr.3: 141292769-141292769 on Build GRCh38
rs370865607 Chr.3: 141292789-141292789 on Build GRCh38
rs779382425 Chr.3: 141292885-141292885 on Build GRCh38
rs755235566 Chr.3: 141292845-141292845 on Build GRCh38
rs777069442 Chr.3: 141292770-141292770 on Build GRCh38
rs376846175 Chr.3: 141292833-141292833 on Build GRCh38
rs754794846 Chr.3: 141292812-141292813 on Build GRCh38
rs781234692 Chr.3: 141292846-141292846 on Build GRCh38
rs780295128 Chr.3: 141292760-141292760 on Build GRCh38
rs145988315 Chr.3: 141292800-141292800 on Build GRCh38
rs200490000 Chr.3: 141292807-141292807 on Build GRCh38
rs931336154 Chr.3: 141292930-141292930 on Build GRCh38
rs3210458 Chr.3: 141292788-141292788 on Build GRCh38
rs745978599 Chr.3: 141292750-141292750 on Build GRCh38
rs753669433 Chr.3: 141292815-141292815 on Build GRCh38
rs758541080 Chr.3: 141292834-141292834 on Build GRCh38
rs369570856 Chr.3: 141292847-141292847 on Build GRCh38
rs915155458 Chr.3: 141292898-141292898 on Build GRCh38
rs750570807 Chr.3: 141292749-141292749 on Build GRCh38
rs748397664 Chr.3: 141292773-141292773 on Build GRCh38
rs138707752 Chr.3: 141292838-141292838 on Build GRCh38

View Full Product Details