Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49886361-49886586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG12
Entrez Gene ID
79087
Gene Name
ALG12, alpha-1,6-mannosyltransferase
Gene Aliases
CDG1G, ECM39, PP14673, hALG12
UniGene
Hs.526711
Gene Chromosome Location
Chr.22: 49860163-49918469 on Build GRCh38
Transcripts
XM_017028936.1 XM_017028937.1 NM_014838.2

Gene Symbol
ZBED4
Entrez Gene ID
9889
Gene Name
zinc finger BED-type containing 4
Gene Aliases
-
UniGene
Hs.475208
Gene Chromosome Location
Chr.22: 49853849-49890078 on Build GRCh38
Transcripts
XM_017028936.1 XM_017028937.1 NM_014838.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3064331 c.2743G>A p.E915K Chr.22: 49886405-49886405 on Build GRCh38
3405728 c.2739C>T p.S913S Chr.22: 49886401-49886401 on Build GRCh38
3064330 c.2740G>A p.V914I Chr.22: 49886402-49886402 on Build GRCh38
3064334 c.2772C>T p.C924C Chr.22: 49886434-49886434 on Build GRCh38
1035266 c.2796C>T p.S932S Chr.22: 49886458-49886458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769364440 Chr.22: 49886434-49886434 on Build GRCh38
rs954512709 Chr.22: 49886511-49886511 on Build GRCh38
rs761274127 Chr.22: 49886428-49886428 on Build GRCh38
rs753270597 Chr.22: 49886475-49886475 on Build GRCh38
rs143483335 Chr.22: 49886470-49886470 on Build GRCh38
rs910312340 Chr.22: 49886558-49886558 on Build GRCh38
rs749555255 Chr.22: 49886405-49886405 on Build GRCh38
rs780912391 Chr.22: 49886498-49886498 on Build GRCh38
rs779100198 Chr.22: 49886491-49886491 on Build GRCh38
rs756539529 Chr.22: 49886482-49886482 on Build GRCh38
rs759215440 Chr.22: 49886540-49886540 on Build GRCh38
rs141714707 Chr.22: 49886495-49886495 on Build GRCh38
rs374917157 Chr.22: 49886458-49886458 on Build GRCh38
rs560097052 Chr.22: 49886508-49886508 on Build GRCh38
rs766859299 Chr.22: 49886380-49886380 on Build GRCh38
rs752866917 Chr.22: 49886401-49886401 on Build GRCh38
rs772839494 Chr.22: 49886506-49886506 on Build GRCh38
rs760174948 Chr.22: 49886435-49886435 on Build GRCh38
rs146661762 Chr.22: 49886392-49886392 on Build GRCh38
rs774127180 Chr.22: 49886518-49886518 on Build GRCh38
rs766048213 Chr.22: 49886459-49886459 on Build GRCh38
rs759298450 Chr.22: 49886469-49886469 on Build GRCh38
rs768644204 Chr.22: 49886416-49886416 on Build GRCh38
rs757531068 Chr.22: 49886490-49886490 on Build GRCh38
rs776515828 Chr.22: 49886418-49886418 on Build GRCh38
rs772533881 Chr.22: 49886437-49886437 on Build GRCh38
rs5770755 Chr.22: 49886488-49886488 on Build GRCh38
rs367851071 Chr.22: 49886509-49886509 on Build GRCh38
rs145947648 Chr.22: 49886496-49886496 on Build GRCh38
rs746792688 Chr.22: 49886410-49886410 on Build GRCh38
rs985784944 Chr.22: 49886557-49886557 on Build GRCh38
rs138867154 Chr.22: 49886501-49886501 on Build GRCh38
rs751918432 Chr.22: 49886385-49886385 on Build GRCh38
rs55681922 Chr.22: 49886402-49886402 on Build GRCh38
rs1042343435 Chr.22: 49886546-49886546 on Build GRCh38
rs761257352 Chr.22: 49886559-49886559 on Build GRCh38
rs775648729 Chr.22: 49886552-49886552 on Build GRCh38
rs779836489 Chr.22: 49886409-49886409 on Build GRCh38
rs371085971 Chr.22: 49886407-49886407 on Build GRCh38
rs755604335 Chr.22: 49886387-49886387 on Build GRCh38
rs1056271900 Chr.22: 49886465-49886465 on Build GRCh38
rs181178469 Chr.22: 49886396-49886396 on Build GRCh38
rs777911902 Chr.22: 49886486-49886486 on Build GRCh38
rs148831653 Chr.22: 49886466-49886466 on Build GRCh38
rs772788326 Chr.22: 49886404-49886404 on Build GRCh38

View Full Product Details