Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38755816-38756089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR5
Entrez Gene ID
79913
Gene Name
ARP5 actin-related protein 5 homolog
Gene Aliases
Arp5, INO80M
UniGene
Hs.371585
Gene Chromosome Location
Chr.20: 38748454-38772446 on Build GRCh38
Transcripts
NM_024855.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4522705 c.1176G>A p.E392E Chr.20: 38756039-38756039 on Build GRCh38
1244195 c.1036C>G p.L346V Chr.20: 38755899-38755899 on Build GRCh38
3546241 c.1017G>A p.M339I Chr.20: 38755880-38755880 on Build GRCh38
7354097 c.1175A>C p.E392A Chr.20: 38756038-38756038 on Build GRCh38
4659173 c.1058C>A p.S353Y Chr.20: 38755921-38755921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991011901 Chr.20: 38756068-38756068 on Build GRCh38
rs775527494 Chr.20: 38755921-38755921 on Build GRCh38
rs775124681 Chr.20: 38756006-38756006 on Build GRCh38
rs1005402315 Chr.20: 38756034-38756034 on Build GRCh38
rs777129616 Chr.20: 38755874-38755874 on Build GRCh38
rs952915218 Chr.20: 38756047-38756048 on Build GRCh38
rs768469393 Chr.20: 38756010-38756010 on Build GRCh38
rs775429264 Chr.20: 38755966-38755966 on Build GRCh38
rs777409273 Chr.20: 38755973-38755973 on Build GRCh38
rs771699094 Chr.20: 38755917-38755917 on Build GRCh38
rs770566142 Chr.20: 38755890-38755890 on Build GRCh38
rs752160959 Chr.20: 38755961-38755961 on Build GRCh38
rs35520380 Chr.20: 38756038-38756038 on Build GRCh38
rs1019135895 Chr.20: 38756001-38756001 on Build GRCh38
rs765339462 Chr.20: 38756030-38756030 on Build GRCh38
rs758867787 Chr.20: 38755955-38755955 on Build GRCh38
rs772679338 Chr.20: 38755977-38755977 on Build GRCh38
rs114985200 Chr.20: 38755991-38755991 on Build GRCh38
rs963764788 Chr.20: 38756067-38756067 on Build GRCh38
rs750612278 Chr.20: 38755948-38755948 on Build GRCh38
rs759341377 Chr.20: 38755907-38755907 on Build GRCh38
rs754022037 Chr.20: 38755979-38755979 on Build GRCh38
rs747007296 Chr.20: 38756009-38756009 on Build GRCh38
rs1011420521 Chr.20: 38755957-38755957 on Build GRCh38
rs376799023 Chr.20: 38755956-38755956 on Build GRCh38
rs888796061 Chr.20: 38755979-38755979 on Build GRCh38
rs748983961 Chr.20: 38755883-38755883 on Build GRCh38
rs1052854057 Chr.20: 38755884-38755884 on Build GRCh38
rs765540329 Chr.20: 38755944-38755944 on Build GRCh38
rs199728746 Chr.20: 38756005-38756005 on Build GRCh38
rs745481440 Chr.20: 38755997-38755997 on Build GRCh38
rs762190046 Chr.20: 38755938-38755938 on Build GRCh38
rs766810765 Chr.20: 38756060-38756060 on Build GRCh38
rs190086095 Chr.20: 38755996-38755996 on Build GRCh38
rs764283343 Chr.20: 38755932-38755932 on Build GRCh38
rs761995847 Chr.20: 38756016-38756016 on Build GRCh38
rs769372168 Chr.20: 38755862-38755862 on Build GRCh38
rs774190205 Chr.20: 38755896-38755896 on Build GRCh38
rs562975557 Chr.20: 38755924-38755924 on Build GRCh38
rs146119497 Chr.20: 38756012-38756012 on Build GRCh38
rs753826278 Chr.20: 38755934-38755934 on Build GRCh38
rs751870107 Chr.20: 38756066-38756066 on Build GRCh38
rs755412578 Chr.20: 38755967-38755967 on Build GRCh38
rs866902970 Chr.20: 38755873-38755873 on Build GRCh38
rs780621580 Chr.20: 38755859-38755859 on Build GRCh38
rs371951076 Chr.20: 38755860-38755860 on Build GRCh38

View Full Product Details