Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36396329-36396554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANLN
Entrez Gene ID
54443
Gene Name
anillin actin binding protein
Gene Aliases
FSGS8, Scraps, scra
UniGene
Hs.62180
Gene Chromosome Location
Chr.7: 36389803-36453791 on Build GRCh38
Transcripts
NM_001284301.2 NM_001284302.2 NM_018685.4 XM_006715746.1 XM_006715747.3 XM_017012354.1 XM_017012355.1 XM_017012356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4931129 c.149A>T p.Q50L Chr.7: 36396396-36396396 on Build GRCh38
5587180 c.96G>A p.R32R Chr.7: 36396343-36396343 on Build GRCh38
1635292 c.115C>T p.R39* Chr.7: 36396362-36396362 on Build GRCh38
1131591 c.98C>G p.S33C Chr.7: 36396345-36396345 on Build GRCh38
4924447 c.116G>T p.R39L Chr.7: 36396363-36396363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779706637 Chr.7: 36396376-36396376 on Build GRCh38
rs149311864 Chr.7: 36396406-36396406 on Build GRCh38
rs202002239 Chr.7: 36396430-36396430 on Build GRCh38
rs751686851 Chr.7: 36396447-36396447 on Build GRCh38
rs778167062 Chr.7: 36396428-36396429 on Build GRCh38
rs764219743 Chr.7: 36396436-36396436 on Build GRCh38
rs753699229 Chr.7: 36396388-36396388 on Build GRCh38
rs976651948 Chr.7: 36396350-36396350 on Build GRCh38
rs759939547 Chr.7: 36396408-36396408 on Build GRCh38
rs761254343 Chr.7: 36396389-36396389 on Build GRCh38
rs35765596 Chr.7: 36396387-36396387 on Build GRCh38
rs992171323 Chr.7: 36396405-36396405 on Build GRCh38
rs767398571 Chr.7: 36396453-36396453 on Build GRCh38
rs138687537 Chr.7: 36396390-36396390 on Build GRCh38
rs1013376515 Chr.7: 36396508-36396508 on Build GRCh38
rs768148237 Chr.7: 36396381-36396381 on Build GRCh38
rs765544159 Chr.7: 36396413-36396413 on Build GRCh38
rs777718099 Chr.7: 36396355-36396355 on Build GRCh38
rs752908729 Chr.7: 36396414-36396414 on Build GRCh38
rs200140060 Chr.7: 36396451-36396451 on Build GRCh38
rs575071809 Chr.7: 36396375-36396375 on Build GRCh38
rs555242135 Chr.7: 36396363-36396363 on Build GRCh38
rs200347027 Chr.7: 36396403-36396403 on Build GRCh38
rs752054050 Chr.7: 36396453-36396453 on Build GRCh38
rs896218853 Chr.7: 36396479-36396479 on Build GRCh38
rs748902962 Chr.7: 36396380-36396380 on Build GRCh38
rs199699357 Chr.7: 36396452-36396453 on Build GRCh38
rs202179258 Chr.7: 36396526-36396526 on Build GRCh38
rs757242868 Chr.7: 36396362-36396362 on Build GRCh38
rs745699628 Chr.7: 36396374-36396374 on Build GRCh38
rs1052128486 Chr.7: 36396456-36396456 on Build GRCh38

View Full Product Details