Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36092329-36092602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRHL2
Entrez Gene ID
54936
Gene Name
ADP-ribosylhydrolase like 2
Gene Aliases
ARH3
UniGene
Hs.18021
Gene Chromosome Location
Chr.1: 36088852-36093932 on Build GRCh38
Transcripts
NM_017825.2 XM_011541636.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1342248 c.717G>T p.E239D Chr.1: 36092437-36092437 on Build GRCh38
5004590 c.771G>A p.S257S Chr.1: 36092491-36092491 on Build GRCh38
4007444 c.733C>T p.R245C Chr.1: 36092453-36092453 on Build GRCh38
1342249 c.732C>T p.S244S Chr.1: 36092452-36092452 on Build GRCh38
5949964 c.711G>T p.M237I Chr.1: 36092431-36092431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991634334 Chr.1: 36092365-36092365 on Build GRCh38
rs376998645 Chr.1: 36092402-36092402 on Build GRCh38
rs374726830 Chr.1: 36092573-36092573 on Build GRCh38
rs369155774 Chr.1: 36092531-36092531 on Build GRCh38
rs765484363 Chr.1: 36092544-36092544 on Build GRCh38
rs764606540 Chr.1: 36092385-36092385 on Build GRCh38
rs780536355 Chr.1: 36092442-36092442 on Build GRCh38
rs776200502 Chr.1: 36092379-36092379 on Build GRCh38
rs370309930 Chr.1: 36092410-36092410 on Build GRCh38
rs866179096 Chr.1: 36092380-36092380 on Build GRCh38
rs1035965112 Chr.1: 36092547-36092547 on Build GRCh38
rs149547220 Chr.1: 36092434-36092434 on Build GRCh38
rs781561741 Chr.1: 36092446-36092446 on Build GRCh38
rs201025691 Chr.1: 36092438-36092438 on Build GRCh38
rs754062810 Chr.1: 36092425-36092425 on Build GRCh38
rs996342477 Chr.1: 36092503-36092503 on Build GRCh38
rs563985546 Chr.1: 36092356-36092356 on Build GRCh38
rs572297139 Chr.1: 36092491-36092491 on Build GRCh38
rs60590162 Chr.1: 36092563-36092563 on Build GRCh38
rs767431801 Chr.1: 36092422-36092422 on Build GRCh38
rs752509406 Chr.1: 36092525-36092525 on Build GRCh38
rs977487565 Chr.1: 36092383-36092383 on Build GRCh38
rs368173504 Chr.1: 36092555-36092555 on Build GRCh38
rs958905882 Chr.1: 36092554-36092554 on Build GRCh38
rs776476743 Chr.1: 36092498-36092498 on Build GRCh38
rs769077572 Chr.1: 36092375-36092375 on Build GRCh38
rs770119014 Chr.1: 36092564-36092564 on Build GRCh38
rs772795321 Chr.1: 36092455-36092455 on Build GRCh38
rs747360712 Chr.1: 36092373-36092373 on Build GRCh38
rs759566942 Chr.1: 36092492-36092492 on Build GRCh38
rs781697208 Chr.1: 36092560-36092560 on Build GRCh38
rs770349925 Chr.1: 36092479-36092479 on Build GRCh38
rs772813149 Chr.1: 36092387-36092387 on Build GRCh38
rs763846082 Chr.1: 36092452-36092452 on Build GRCh38
rs762515954 Chr.1: 36092466-36092466 on Build GRCh38
rs769460369 Chr.1: 36092454-36092454 on Build GRCh38
rs924589061 Chr.1: 36092388-36092388 on Build GRCh38
rs376099826 Chr.1: 36092381-36092381 on Build GRCh38
rs114293583 Chr.1: 36092370-36092370 on Build GRCh38
rs375280387 Chr.1: 36092419-36092419 on Build GRCh38
rs372033314 Chr.1: 36092559-36092559 on Build GRCh38
rs577682830 Chr.1: 36092371-36092371 on Build GRCh38
rs758438678 Chr.1: 36092553-36092553 on Build GRCh38
rs757530458 Chr.1: 36092430-36092430 on Build GRCh38
rs372899778 Chr.1: 36092572-36092572 on Build GRCh38
rs890121750 Chr.1: 36092548-36092548 on Build GRCh38
rs752761171 Chr.1: 36092404-36092404 on Build GRCh38
rs777415357 Chr.1: 36092567-36092567 on Build GRCh38
rs752016206 Chr.1: 36092557-36092557 on Build GRCh38
rs750678843 Chr.1: 36092549-36092549 on Build GRCh38
rs747763140 Chr.1: 36092447-36092447 on Build GRCh38
rs747520525 Chr.1: 36092443-36092443 on Build GRCh38
rs914663818 Chr.1: 36092369-36092369 on Build GRCh38
rs754749794 Chr.1: 36092417-36092417 on Build GRCh38
rs1003181405 Chr.1: 36092540-36092540 on Build GRCh38
rs767378654 Chr.1: 36092512-36092512 on Build GRCh38
rs766322358 Chr.1: 36092403-36092403 on Build GRCh38
rs148612837 Chr.1: 36092439-36092439 on Build GRCh38

View Full Product Details