Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102483961-102484214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR1A
Entrez Gene ID
10121
Gene Name
ARP1 actin-related protein 1 homolog A, centractin alpha
Gene Aliases
ARP1, Arp1A, CTRN1
UniGene
Hs.153961
Gene Chromosome Location
Chr.10: 102479229-102502755 on Build GRCh38
Transcripts
NM_005736.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2059370 c.620C>T p.S207L Chr.10: 102484197-102484197 on Build GRCh38
6576953 c.642C>G p.V214V Chr.10: 102484175-102484175 on Build GRCh38
4779100 c.655G>C p.E219Q Chr.10: 102484162-102484162 on Build GRCh38
5485357 c.647C>T p.A216V Chr.10: 102484170-102484170 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375743127 Chr.10: 102484139-102484139 on Build GRCh38
rs933927872 Chr.10: 102484068-102484068 on Build GRCh38
rs780809653 Chr.10: 102484124-102484124 on Build GRCh38
rs370101460 Chr.10: 102484146-102484146 on Build GRCh38
rs976102990 Chr.10: 102484040-102484040 on Build GRCh38
rs748488545 Chr.10: 102484079-102484079 on Build GRCh38
rs775283413 Chr.10: 102484027-102484027 on Build GRCh38
rs539085682 Chr.10: 102484013-102484013 on Build GRCh38
rs985312778 Chr.10: 102484069-102484069 on Build GRCh38
rs918603967 Chr.10: 102484108-102484108 on Build GRCh38
rs998336106 Chr.10: 102483983-102483983 on Build GRCh38
rs7079810 Chr.10: 102484126-102484126 on Build GRCh38
rs182197890 Chr.10: 102484147-102484147 on Build GRCh38
rs767490090 Chr.10: 102484150-102484150 on Build GRCh38
rs935029284 Chr.10: 102484148-102484148 on Build GRCh38
rs962383593 Chr.10: 102484032-102484032 on Build GRCh38
rs1046446074 Chr.10: 102484103-102484103 on Build GRCh38
rs925499141 Chr.10: 102484048-102484048 on Build GRCh38
rs958447879 Chr.10: 102484058-102484058 on Build GRCh38
rs766935547 Chr.10: 102484189-102484189 on Build GRCh38
rs779162172 Chr.10: 102484186-102484186 on Build GRCh38
rs769675285 Chr.10: 102483984-102483984 on Build GRCh38
rs949523102 Chr.10: 102484100-102484100 on Build GRCh38
rs761032920 Chr.10: 102484185-102484185 on Build GRCh38
rs775366848 Chr.10: 102484143-102484143 on Build GRCh38
rs550508462 Chr.10: 102484066-102484066 on Build GRCh38
rs984892064 Chr.10: 102484012-102484012 on Build GRCh38
rs1019991298 Chr.10: 102483996-102483996 on Build GRCh38
rs991041220 Chr.10: 102484059-102484059 on Build GRCh38
rs200290108 Chr.10: 102484173-102484173 on Build GRCh38
rs375214232 Chr.10: 102484166-102484166 on Build GRCh38
rs879088927 Chr.10: 102484092-102484092 on Build GRCh38

View Full Product Details