Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 42216099-42216368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG1
Entrez Gene ID
9619
Gene Name
ATP binding cassette subfamily G member 1
Gene Aliases
ABC8, WHITE1
UniGene
Hs.124649
Gene Chromosome Location
Chr.21: 42199689-42304389 on Build GRCh38
Transcripts
NM_207627.1 NM_207628.1 NM_207629.1

SNP Detail

SNP ID Chromosome Location
rs997651405 Chr.21: 42216150-42216150 on Build GRCh38
rs1036456933 Chr.21: 42216293-42216293 on Build GRCh38
rs997703600 Chr.21: 42216322-42216322 on Build GRCh38
rs532689435 Chr.21: 42216313-42216313 on Build GRCh38
rs1021739861 Chr.21: 42216162-42216162 on Build GRCh38
rs968396684 Chr.21: 42216163-42216163 on Build GRCh38
rs142204098 Chr.21: 42216136-42216136 on Build GRCh38
rs8126971 Chr.21: 42216302-42216302 on Build GRCh38
rs572638249 Chr.21: 42216237-42216237 on Build GRCh38
rs537021695 Chr.21: 42216170-42216170 on Build GRCh38
rs112156022 Chr.21: 42216124-42216124 on Build GRCh38
rs985829229 Chr.21: 42216160-42216160 on Build GRCh38
rs543818244 Chr.21: 42216291-42216291 on Build GRCh38
rs7275664 Chr.21: 42216347-42216347 on Build GRCh38
rs546305232 Chr.21: 42216179-42216179 on Build GRCh38
rs8129302 Chr.21: 42216213-42216213 on Build GRCh38
rs575214231 Chr.21: 42216252-42216252 on Build GRCh38
rs753569532 Chr.21: 42216182-42216182 on Build GRCh38
rs759270699 Chr.21: 42216138-42216138 on Build GRCh38
rs1045222839 Chr.21: 42216197-42216197 on Build GRCh38
rs923907961 Chr.21: 42216141-42216141 on Build GRCh38
rs181861233 Chr.21: 42216209-42216209 on Build GRCh38
rs956698162 Chr.21: 42216152-42216152 on Build GRCh38
rs956646674 Chr.21: 42216333-42216333 on Build GRCh38
rs992151725 Chr.21: 42216338-42216338 on Build GRCh38
rs537842530 Chr.21: 42216247-42216247 on Build GRCh38
rs568441907 Chr.21: 42216153-42216153 on Build GRCh38
rs868789254 Chr.21: 42216273-42216273 on Build GRCh38
rs149713099 Chr.21: 42216196-42216196 on Build GRCh38
rs1030924554 Chr.21: 42216319-42216319 on Build GRCh38
rs1047776621 Chr.21: 42216214-42216214 on Build GRCh38
rs541504673 Chr.21: 42216241-42216241 on Build GRCh38
rs764826860 Chr.21: 42216140-42216140 on Build GRCh38
rs34068794 Chr.21: 42216141-42216142 on Build GRCh38
rs751643895 Chr.21: 42216332-42216332 on Build GRCh38
rs942038607 Chr.21: 42216251-42216251 on Build GRCh38
rs570353812 Chr.21: 42216246-42216246 on Build GRCh38
rs751557147 Chr.21: 42216187-42216187 on Build GRCh38

View Full Product Details