Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36011305-36011574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH6
Entrez Gene ID
84964
Gene Name
alkB homolog 6
Gene Aliases
ABH6
UniGene
Hs.71941
Gene Chromosome Location
Chr.19: 36009116-36014243 on Build GRCh38
Transcripts
NM_001297701.1 NM_032878.3 NM_198867.1 XM_005259357.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6465709 c.213T>A p.F71L Chr.19: 36011459-36011459 on Build GRCh38
6465708 c.214A>T p.N72Y Chr.19: 36011458-36011458 on Build GRCh38
3199337 c.211T>G p.F71V Chr.19: 36011461-36011461 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs940870840 Chr.19: 36011377-36011377 on Build GRCh38
rs755392260 Chr.19: 36011479-36011479 on Build GRCh38
rs750471811 Chr.19: 36011538-36011538 on Build GRCh38
rs753225911 Chr.19: 36011489-36011489 on Build GRCh38
rs747014083 Chr.19: 36011511-36011511 on Build GRCh38
rs973575491 Chr.19: 36011467-36011468 on Build GRCh38
rs765019035 Chr.19: 36011386-36011386 on Build GRCh38
rs775488775 Chr.19: 36011394-36011394 on Build GRCh38
rs866485651 Chr.19: 36011438-36011438 on Build GRCh38
rs571639661 Chr.19: 36011496-36011496 on Build GRCh38
rs373876497 Chr.19: 36011500-36011500 on Build GRCh38
rs763637367 Chr.19: 36011408-36011408 on Build GRCh38
rs986999903 Chr.19: 36011381-36011381 on Build GRCh38
rs781298307 Chr.19: 36011460-36011460 on Build GRCh38
rs771015590 Chr.19: 36011513-36011513 on Build GRCh38
rs759265857 Chr.19: 36011376-36011376 on Build GRCh38
rs771103863 Chr.19: 36011367-36011367 on Build GRCh38
rs776863753 Chr.19: 36011368-36011368 on Build GRCh38
rs987977749 Chr.19: 36011329-36011329 on Build GRCh38
rs762852227 Chr.19: 36011397-36011397 on Build GRCh38
rs200768994 Chr.19: 36011427-36011427 on Build GRCh38
rs756857341 Chr.19: 36011450-36011450 on Build GRCh38
rs192706685 Chr.19: 36011504-36011504 on Build GRCh38
rs773218077 Chr.19: 36011365-36011365 on Build GRCh38
rs750105338 Chr.19: 36011471-36011471 on Build GRCh38
rs767229034 Chr.19: 36011457-36011457 on Build GRCh38
rs779478727 Chr.19: 36011484-36011484 on Build GRCh38
rs8111422 Chr.19: 36011539-36011539 on Build GRCh38

View Full Product Details