Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32171264-32171533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT1
Entrez Gene ID
10554
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases
1-AGPAT1, G15, LPAAT-alpha, LPAATA
UniGene
Hs.409230
Gene Chromosome Location
Chr.6: 32168206-32178111 on Build GRCh38
Transcripts
NM_006411.3 NM_032741.4 XM_005248805.3 XM_005248806.2 XM_011514234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
39812 c.69C>T p.P23P Chr.6: 32171428-32171428 on Build GRCh38
5870296 c.86G>A p.S29N Chr.6: 32171411-32171411 on Build GRCh38
252330 c.128G>A p.G43D Chr.6: 32171369-32171369 on Build GRCh38
252329 c.190G>A p.E64K Chr.6: 32171307-32171307 on Build GRCh38
2150668 c.181C>T p.R61C Chr.6: 32171316-32171316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765047623 Chr.6: 32171501-32171501 on Build GRCh38
rs754930631 Chr.6: 32171430-32171444 on Build GRCh38
rs780487704 Chr.6: 32171358-32171358 on Build GRCh38
rs749653275 Chr.6: 32171388-32171388 on Build GRCh38
rs756370870 Chr.6: 32171356-32171356 on Build GRCh38
rs529759736 Chr.6: 32171484-32171484 on Build GRCh38
rs772759702 Chr.6: 32171424-32171424 on Build GRCh38
rs977923338 Chr.6: 32171456-32171456 on Build GRCh38
rs771437202 Chr.6: 32171422-32171422 on Build GRCh38
rs750516488 Chr.6: 32171340-32171340 on Build GRCh38
rs1010947497 Chr.6: 32171314-32171314 on Build GRCh38
rs770568102 Chr.6: 32171434-32171434 on Build GRCh38
rs200978599 Chr.6: 32171310-32171310 on Build GRCh38
rs765147808 Chr.6: 32171307-32171307 on Build GRCh38
rs777983200 Chr.6: 32171508-32171509 on Build GRCh38
rs759269879 Chr.6: 32171466-32171466 on Build GRCh38
rs752527527 Chr.6: 32171456-32171458 on Build GRCh38
rs769152569 Chr.6: 32171406-32171406 on Build GRCh38
rs779060823 Chr.6: 32171432-32171432 on Build GRCh38
rs760144285 Chr.6: 32171426-32171426 on Build GRCh38
rs776323158 Chr.6: 32171451-32171451 on Build GRCh38
rs547166818 Chr.6: 32171325-32171325 on Build GRCh38
rs756914433 Chr.6: 32171381-32171381 on Build GRCh38
rs758140924 Chr.6: 32171455-32171456 on Build GRCh38
rs757244240 Chr.6: 32171311-32171311 on Build GRCh38
rs966529131 Chr.6: 32171455-32171455 on Build GRCh38
rs11964847 Chr.6: 32171409-32171409 on Build GRCh38

View Full Product Details