Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 8937968-8938197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCL3
Entrez Gene ID
56676
Gene Name
achaete-scute family bHLH transcription factor 3
Gene Aliases
HASH3, SGN1, bHLHa42
UniGene
Hs.501852
Gene Chromosome Location
Chr.11: 8937572-8943033 on Build GRCh38
Transcripts
NM_020646.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5031279 c.191T>C p.I64T Chr.11: 8937971-8937971 on Build GRCh38
4633455 c.26G>T p.S9I Chr.11: 8938136-8938136 on Build GRCh38
2222593 c.91C>T p.L31L Chr.11: 8938071-8938071 on Build GRCh38
5613474 c.164G>C p.R55P Chr.11: 8937998-8937998 on Build GRCh38
7417666 c.127G>T p.A43S Chr.11: 8938035-8938035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025835100 Chr.11: 8938007-8938007 on Build GRCh38
rs934269240 Chr.11: 8938061-8938061 on Build GRCh38
rs761961023 Chr.11: 8938006-8938006 on Build GRCh38
rs763131471 Chr.11: 8938084-8938084 on Build GRCh38
rs36020286 Chr.11: 8938097-8938097 on Build GRCh38
rs754085759 Chr.11: 8938065-8938065 on Build GRCh38
rs191792614 Chr.11: 8937999-8937999 on Build GRCh38
rs781435579 Chr.11: 8938034-8938034 on Build GRCh38
rs745498896 Chr.11: 8938056-8938056 on Build GRCh38
rs186562703 Chr.11: 8937993-8937993 on Build GRCh38
rs766480799 Chr.11: 8938091-8938091 on Build GRCh38
rs377109433 Chr.11: 8938023-8938023 on Build GRCh38
rs145370594 Chr.11: 8938113-8938113 on Build GRCh38
rs769670995 Chr.11: 8938157-8938157 on Build GRCh38
rs781678039 Chr.11: 8938107-8938107 on Build GRCh38
rs753627385 Chr.11: 8938001-8938001 on Build GRCh38
rs750694268 Chr.11: 8938082-8938082 on Build GRCh38
rs761737334 Chr.11: 8938077-8938077 on Build GRCh38
rs1009296563 Chr.11: 8938116-8938116 on Build GRCh38
rs765434725 Chr.11: 8938018-8938018 on Build GRCh38
rs35338189 Chr.11: 8938016-8938016 on Build GRCh38
rs770571296 Chr.11: 8937989-8937989 on Build GRCh38
rs771700039 Chr.11: 8937996-8937996 on Build GRCh38
rs202112150 Chr.11: 8938075-8938075 on Build GRCh38
rs756660185 Chr.11: 8938109-8938109 on Build GRCh38
rs775108865 Chr.11: 8938062-8938062 on Build GRCh38
rs199698276 Chr.11: 8938031-8938031 on Build GRCh38
rs150048631 Chr.11: 8938072-8938072 on Build GRCh38
rs764929260 Chr.11: 8938078-8938078 on Build GRCh38
rs768048442 Chr.11: 8938136-8938136 on Build GRCh38
rs776690894 Chr.11: 8938139-8938139 on Build GRCh38
rs1037013452 Chr.11: 8938045-8938045 on Build GRCh38
rs755896237 Chr.11: 8938041-8938041 on Build GRCh38
rs777974131 Chr.11: 8938115-8938115 on Build GRCh38
rs144588221 Chr.11: 8938048-8938048 on Build GRCh38
rs937801002 Chr.11: 8938009-8938009 on Build GRCh38
rs183622183 Chr.11: 8938150-8938150 on Build GRCh38
rs200189639 Chr.11: 8938030-8938030 on Build GRCh38
rs533929038 Chr.11: 8938134-8938134 on Build GRCh38
rs4909951 Chr.11: 8937998-8937998 on Build GRCh38
rs375148614 Chr.11: 8937991-8937991 on Build GRCh38
rs748489069 Chr.11: 8938044-8938044 on Build GRCh38
rs745555998 Chr.11: 8938119-8938119 on Build GRCh38
rs770491181 Chr.11: 8938047-8938047 on Build GRCh38
rs202151573 Chr.11: 8938098-8938098 on Build GRCh38
rs750904681 Chr.11: 8938029-8938029 on Build GRCh38
rs753137271 Chr.11: 8938111-8938111 on Build GRCh38
rs35625375 Chr.11: 8938134-8938135 on Build GRCh38
rs758083524 Chr.11: 8938121-8938121 on Build GRCh38
rs201803232 Chr.11: 8938060-8938060 on Build GRCh38

View Full Product Details