Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50790821-50791056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACPT
Entrez Gene ID
93650
Gene Name
acid phosphatase, testicular
Gene Aliases
-
UniGene
Hs.293394
Gene Chromosome Location
Chr.19: 50790415-50795224 on Build GRCh38
Transcripts
NM_033068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354005 c.293G>T p.R98L Chr.19: 50790850-50790850 on Build GRCh38
4132328 c.267C>T p.Y89Y Chr.19: 50790824-50790824 on Build GRCh38
7354006 c.303G>A p.E101E Chr.19: 50790860-50790860 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs567340920 Chr.19: 50791002-50791002 on Build GRCh38
rs1036553797 Chr.19: 50790980-50790980 on Build GRCh38
rs954474680 Chr.19: 50790894-50790894 on Build GRCh38
rs909307758 Chr.19: 50790925-50790925 on Build GRCh38
rs777345491 Chr.19: 50790876-50790876 on Build GRCh38
rs1046487233 Chr.19: 50790999-50790999 on Build GRCh38
rs556936186 Chr.19: 50790998-50790998 on Build GRCh38
rs868158209 Chr.19: 50790862-50790862 on Build GRCh38
rs992164531 Chr.19: 50790872-50790872 on Build GRCh38
rs973257747 Chr.19: 50790966-50790966 on Build GRCh38
rs755504814 Chr.19: 50791005-50791005 on Build GRCh38
rs750759139 Chr.19: 50790892-50790892 on Build GRCh38
rs185153035 Chr.19: 50790842-50790842 on Build GRCh38
rs916607166 Chr.19: 50790881-50790881 on Build GRCh38
rs752416597 Chr.19: 50790853-50790853 on Build GRCh38
rs145475625 Chr.19: 50790841-50790841 on Build GRCh38
rs188470865 Chr.19: 50790995-50790995 on Build GRCh38
rs940814045 Chr.19: 50790977-50790977 on Build GRCh38
rs765700801 Chr.19: 50790870-50790870 on Build GRCh38
rs910385164 Chr.19: 50790935-50790935 on Build GRCh38
rs375408855 Chr.19: 50790850-50790850 on Build GRCh38
rs200279792 Chr.19: 50790845-50790845 on Build GRCh38
rs902080998 Chr.19: 50790988-50790988 on Build GRCh38
rs977908427 Chr.19: 50790852-50790852 on Build GRCh38
rs923836610 Chr.19: 50790859-50790859 on Build GRCh38
rs372492348 Chr.19: 50790904-50790904 on Build GRCh38
rs966996662 Chr.19: 50790878-50790878 on Build GRCh38
rs375922906 Chr.19: 50790953-50790953 on Build GRCh38
rs963300996 Chr.19: 50790941-50790941 on Build GRCh38
rs978558637 Chr.19: 50790856-50790856 on Build GRCh38
rs577676009 Chr.19: 50790882-50790882 on Build GRCh38
rs200227680 Chr.19: 50790866-50790866 on Build GRCh38
rs115294146 Chr.19: 50791003-50791003 on Build GRCh38

View Full Product Details