Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49884053-49884308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG12
Entrez Gene ID
79087
Gene Name
ALG12, alpha-1,6-mannosyltransferase
Gene Aliases
CDG1G, ECM39, PP14673, hALG12
UniGene
Hs.526711
Gene Chromosome Location
Chr.22: 49860163-49918469 on Build GRCh38
Transcripts
XM_017028936.1 XM_017028937.1 NM_014838.2

Gene Symbol
ZBED4
Entrez Gene ID
9889
Gene Name
zinc finger BED-type containing 4
Gene Aliases
-
UniGene
Hs.475208
Gene Chromosome Location
Chr.22: 49853849-49890078 on Build GRCh38
Transcripts
XM_017028936.1 XM_017028937.1 NM_014838.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5726731 c.461C>T p.T154I Chr.22: 49884123-49884123 on Build GRCh38
3842899 c.483G>A p.V161V Chr.22: 49884145-49884145 on Build GRCh38
3800273 c.498G>A p.P166P Chr.22: 49884160-49884160 on Build GRCh38
5441699 c.600C>T p.S200S Chr.22: 49884262-49884262 on Build GRCh38
5463137 c.394A>G p.T132A Chr.22: 49884056-49884056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112831203 Chr.22: 49884106-49884106 on Build GRCh38
rs768717087 Chr.22: 49884166-49884166 on Build GRCh38
rs777914925 Chr.22: 49884150-49884150 on Build GRCh38
rs118101222 Chr.22: 49884250-49884250 on Build GRCh38
rs201499212 Chr.22: 49884142-49884142 on Build GRCh38
rs780657401 Chr.22: 49884285-49884285 on Build GRCh38
rs146482955 Chr.22: 49884205-49884205 on Build GRCh38
rs760796980 Chr.22: 49884218-49884218 on Build GRCh38
rs372789246 Chr.22: 49884277-49884277 on Build GRCh38
rs747715026 Chr.22: 49884172-49884172 on Build GRCh38
rs142948522 Chr.22: 49884249-49884249 on Build GRCh38
rs577572441 Chr.22: 49884268-49884268 on Build GRCh38
rs1049332809 Chr.22: 49884149-49884149 on Build GRCh38
rs868834816 Chr.22: 49884143-49884143 on Build GRCh38
rs761279456 Chr.22: 49884284-49884284 on Build GRCh38
rs781323630 Chr.22: 49884237-49884237 on Build GRCh38
rs144068518 Chr.22: 49884196-49884196 on Build GRCh38
rs772779249 Chr.22: 49884188-49884188 on Build GRCh38
rs766733274 Chr.22: 49884088-49884088 on Build GRCh38
rs768079765 Chr.22: 49884199-49884199 on Build GRCh38
rs757524342 Chr.22: 49884226-49884226 on Build GRCh38
rs891795677 Chr.22: 49884181-49884181 on Build GRCh38
rs753189667 Chr.22: 49884204-49884204 on Build GRCh38
rs758171600 Chr.22: 49884157-49884157 on Build GRCh38
rs908397289 Chr.22: 49884214-49884214 on Build GRCh38
rs778933183 Chr.22: 49884228-49884228 on Build GRCh38
rs367826109 Chr.22: 49884243-49884243 on Build GRCh38
rs751463067 Chr.22: 49884234-49884234 on Build GRCh38
rs910796 Chr.22: 49884187-49884187 on Build GRCh38
rs148606148 Chr.22: 49884151-49884151 on Build GRCh38
rs777363455 Chr.22: 49884247-49884247 on Build GRCh38
rs765045341 Chr.22: 49884080-49884080 on Build GRCh38
rs76232326 Chr.22: 49884165-49884165 on Build GRCh38
rs1031740512 Chr.22: 49884171-49884171 on Build GRCh38
rs139936208 Chr.22: 49884220-49884220 on Build GRCh38
rs767956723 Chr.22: 49884124-49884124 on Build GRCh38
rs949003382 Chr.22: 49884152-49884152 on Build GRCh38
rs148554740 Chr.22: 49884244-49884244 on Build GRCh38
rs752780494 Chr.22: 49884136-49884136 on Build GRCh38
rs377741840 Chr.22: 49884190-49884190 on Build GRCh38
rs766228106 Chr.22: 49884224-49884226 on Build GRCh38
rs781267128 Chr.22: 49884169-49884169 on Build GRCh38
rs374708587 Chr.22: 49884197-49884197 on Build GRCh38
rs751828813 Chr.22: 49884100-49884100 on Build GRCh38
rs141158718 Chr.22: 49884160-49884160 on Build GRCh38
rs776021437 Chr.22: 49884281-49884281 on Build GRCh38
rs772131553 Chr.22: 49884085-49884085 on Build GRCh38
rs759952594 Chr.22: 49884266-49884266 on Build GRCh38
rs772902530 Chr.22: 49884084-49884084 on Build GRCh38
rs150734381 Chr.22: 49884164-49884164 on Build GRCh38
rs910797 Chr.22: 49884235-49884235 on Build GRCh38
rs199553823 Chr.22: 49884248-49884248 on Build GRCh38

View Full Product Details