Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150983898-150984145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA9
Entrez Gene ID
8416
Gene Name
annexin A9
Gene Aliases
ANX31
UniGene
Hs.3346
Gene Chromosome Location
Chr.1: 150982023-150995638 on Build GRCh38
Transcripts
NM_003568.2 XM_011510058.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2182857 c.199G>A p.V67M Chr.1: 150984001-150984001 on Build GRCh38
3473546 c.249C>T p.F83F Chr.1: 150984051-150984051 on Build GRCh38
349762 c.218G>C p.R73T Chr.1: 150984020-150984020 on Build GRCh38
4479126 c.229C>A p.Q77K Chr.1: 150984031-150984031 on Build GRCh38
4773355 c.247T>G p.F83V Chr.1: 150984049-150984049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747250822 Chr.1: 150983968-150983968 on Build GRCh38
rs372342798 Chr.1: 150984101-150984101 on Build GRCh38
rs769830440 Chr.1: 150984043-150984043 on Build GRCh38
rs758360234 Chr.1: 150984096-150984096 on Build GRCh38
rs748360796 Chr.1: 150984117-150984117 on Build GRCh38
rs1050239009 Chr.1: 150983950-150983950 on Build GRCh38
rs998784689 Chr.1: 150983980-150983980 on Build GRCh38
rs764836508 Chr.1: 150984083-150984083 on Build GRCh38
rs748928871 Chr.1: 150983972-150983982 on Build GRCh38
rs374447843 Chr.1: 150984104-150984104 on Build GRCh38
rs766757747 Chr.1: 150983992-150983992 on Build GRCh38
rs761839467 Chr.1: 150983977-150983977 on Build GRCh38
rs368497566 Chr.1: 150984014-150984014 on Build GRCh38
rs757267954 Chr.1: 150983926-150983926 on Build GRCh38
rs765316963 Chr.1: 150984003-150984003 on Build GRCh38
rs587737229 Chr.1: 150983969-150983969 on Build GRCh38
rs1055713939 Chr.1: 150984119-150984119 on Build GRCh38
rs182619039 Chr.1: 150984038-150984038 on Build GRCh38
rs781736223 Chr.1: 150983967-150983967 on Build GRCh38
rs147042782 Chr.1: 150984059-150984059 on Build GRCh38
rs761450893 Chr.1: 150984068-150984068 on Build GRCh38
rs750354694 Chr.1: 150984013-150984013 on Build GRCh38
rs745681041 Chr.1: 150983956-150983956 on Build GRCh38
rs372750432 Chr.1: 150984031-150984031 on Build GRCh38
rs201382965 Chr.1: 150984058-150984058 on Build GRCh38
rs888991007 Chr.1: 150983960-150983960 on Build GRCh38
rs781606648 Chr.1: 150983925-150983925 on Build GRCh38
rs760392703 Chr.1: 150983995-150983995 on Build GRCh38
rs933051281 Chr.1: 150984087-150984087 on Build GRCh38
rs776695364 Chr.1: 150983976-150983976 on Build GRCh38
rs750444420 Chr.1: 150984092-150984092 on Build GRCh38
rs771879546 Chr.1: 150983963-150983963 on Build GRCh38
rs763262952 Chr.1: 150983986-150983986 on Build GRCh38
rs369682716 Chr.1: 150984107-150984107 on Build GRCh38
rs138047919 Chr.1: 150984053-150984053 on Build GRCh38
rs773683908 Chr.1: 150983984-150983984 on Build GRCh38
rs970969708 Chr.1: 150983918-150983918 on Build GRCh38
rs138556601 Chr.1: 150984023-150984023 on Build GRCh38
rs779911311 Chr.1: 150984020-150984020 on Build GRCh38
rs142629423 Chr.1: 150984054-150984054 on Build GRCh38
rs936298285 Chr.1: 150984115-150984115 on Build GRCh38
rs763589794 Chr.1: 150984000-150984000 on Build GRCh38
rs781430856 Chr.1: 150984109-150984109 on Build GRCh38
rs143089777 Chr.1: 150983993-150983993 on Build GRCh38
rs192303395 Chr.1: 150983932-150983932 on Build GRCh38
rs781341084 Chr.1: 150984034-150984034 on Build GRCh38
rs753432922 Chr.1: 150984001-150984001 on Build GRCh38
rs746398932 Chr.1: 150983983-150983983 on Build GRCh38
rs113700907 Chr.1: 150984033-150984033 on Build GRCh38
rs763864001 Chr.1: 150984063-150984063 on Build GRCh38
rs758279562 Chr.1: 150983961-150983961 on Build GRCh38

View Full Product Details