Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47168032-47168287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGAP2
Entrez Gene ID
84364
Gene Name
ADP ribosylation factor GTPase activating protein 2
Gene Aliases
IRZ, NBLA10535, ZFP289, ZNF289
UniGene
Hs.436204
Gene Chromosome Location
Chr.11: 47164298-47177503 on Build GRCh38
Transcripts
NM_001242832.1 NM_032389.4 XM_005253166.1 XM_005253167.1 XM_005253168.2 XM_006718346.1 XM_011520405.1 XM_017018413.1 XM_017018414.1 XM_017018415.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
927359 c.994C>T p.P332S Chr.11: 47168199-47168199 on Build GRCh38
927358 c.1081A>G p.N361D Chr.11: 47168033-47168033 on Build GRCh38
4574390 c.1038C>T p.D346D Chr.11: 47168155-47168155 on Build GRCh38
272349 c.957C>T p.S319S Chr.11: 47168236-47168236 on Build GRCh38
7095631 c.1071G>C p.K357N Chr.11: 47168043-47168043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557889655 Chr.11: 47168155-47168155 on Build GRCh38
rs762208978 Chr.11: 47168219-47168219 on Build GRCh38
rs754848729 Chr.11: 47168170-47168170 on Build GRCh38
rs1054325726 Chr.11: 47168137-47168137 on Build GRCh38
rs369801651 Chr.11: 47168128-47168128 on Build GRCh38
rs779066303 Chr.11: 47168113-47168113 on Build GRCh38
rs747100346 Chr.11: 47168202-47168202 on Build GRCh38
rs751052486 Chr.11: 47168234-47168234 on Build GRCh38
rs766032167 Chr.11: 47168232-47168232 on Build GRCh38
rs1042416778 Chr.11: 47168122-47168122 on Build GRCh38
rs376762616 Chr.11: 47168109-47168109 on Build GRCh38
rs747947634 Chr.11: 47168086-47168086 on Build GRCh38
rs759108635 Chr.11: 47168237-47168237 on Build GRCh38
rs780897773 Chr.11: 47168071-47168071 on Build GRCh38
rs965264850 Chr.11: 47168254-47168254 on Build GRCh38
rs772459221 Chr.11: 47168183-47168183 on Build GRCh38
rs572840473 Chr.11: 47168105-47168105 on Build GRCh38
rs762144749 Chr.11: 47168152-47168152 on Build GRCh38
rs200484551 Chr.11: 47168255-47168255 on Build GRCh38
rs373373969 Chr.11: 47168147-47168147 on Build GRCh38
rs752530691 Chr.11: 47168174-47168174 on Build GRCh38
rs770003867 Chr.11: 47168225-47168225 on Build GRCh38
rs375786607 Chr.11: 47168230-47168230 on Build GRCh38
rs761227357 Chr.11: 47168135-47168135 on Build GRCh38
rs557366591 Chr.11: 47168176-47168176 on Build GRCh38
rs767408649 Chr.11: 47168248-47168248 on Build GRCh38
rs762445464 Chr.11: 47168231-47168231 on Build GRCh38
rs963472806 Chr.11: 47168240-47168240 on Build GRCh38
rs892874570 Chr.11: 47168062-47168062 on Build GRCh38
rs777079416 Chr.11: 47168143-47168143 on Build GRCh38
rs769293804 Chr.11: 47168140-47168140 on Build GRCh38
rs146816848 Chr.11: 47168180-47168180 on Build GRCh38
rs34662994 Chr.11: 47168177-47168177 on Build GRCh38
rs372979011 Chr.11: 47168256-47168256 on Build GRCh38
rs746031015 Chr.11: 47168178-47168178 on Build GRCh38
rs766721530 Chr.11: 47168058-47168058 on Build GRCh38
rs781160679 Chr.11: 47168173-47168173 on Build GRCh38
rs902867332 Chr.11: 47168125-47168125 on Build GRCh38
rs751073020 Chr.11: 47168169-47168169 on Build GRCh38
rs771168283 Chr.11: 47168088-47168088 on Build GRCh38
rs745925827 Chr.11: 47168117-47168117 on Build GRCh38
rs775738879 Chr.11: 47168130-47168130 on Build GRCh38
rs780219540 Chr.11: 47168184-47168184 on Build GRCh38
rs769273203 Chr.11: 47168216-47168216 on Build GRCh38
rs777346898 Chr.11: 47168218-47168218 on Build GRCh38
rs754937717 Chr.11: 47168066-47168066 on Build GRCh38
rs1011764172 Chr.11: 47168116-47168116 on Build GRCh38
rs373135315 Chr.11: 47168235-47168235 on Build GRCh38
rs1017793859 Chr.11: 47168236-47168236 on Build GRCh38
rs545061391 Chr.11: 47168262-47168262 on Build GRCh38
rs752622382 Chr.11: 47168249-47168249 on Build GRCh38
rs764939758 Chr.11: 47168154-47168154 on Build GRCh38
rs544068883 Chr.11: 47168161-47168161 on Build GRCh38
rs919231600 Chr.11: 47168267-47168267 on Build GRCh38
rs369659958 Chr.11: 47168157-47168157 on Build GRCh38

View Full Product Details