Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42852546-42852777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOC3
Entrez Gene ID
8639
Gene Name
amine oxidase, copper containing 3
Gene Aliases
HPAO, SSAO, VAP-1, VAP1
UniGene
Hs.198241
Gene Chromosome Location
Chr.17: 42851123-42858130 on Build GRCh38
Transcripts
NM_001277731.1 NM_003734.3 NR_102422.1 XM_011525419.1 XM_011525420.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
42799 c.1361G>A p.G454D Chr.17: 42852704-42852704 on Build GRCh38
3517824 c.1245C>T p.F415F Chr.17: 42852588-42852588 on Build GRCh38
174848 c.1347G>A p.S449S Chr.17: 42852690-42852690 on Build GRCh38
3517825 c.1383C>T p.V461V Chr.17: 42852726-42852726 on Build GRCh38
3517823 c.1233G>A p.V411V Chr.17: 42852576-42852576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759731358 Chr.17: 42852660-42852660 on Build GRCh38
rs113316620 Chr.17: 42852634-42852634 on Build GRCh38
rs775361075 Chr.17: 42852573-42852573 on Build GRCh38
rs200247368 Chr.17: 42852644-42852644 on Build GRCh38
rs369071538 Chr.17: 42852665-42852665 on Build GRCh38
rs146228333 Chr.17: 42852654-42852654 on Build GRCh38
rs769259655 Chr.17: 42852622-42852622 on Build GRCh38
rs557853983 Chr.17: 42852655-42852655 on Build GRCh38
rs770066884 Chr.17: 42852738-42852738 on Build GRCh38
rs773361574 Chr.17: 42852583-42852583 on Build GRCh38
rs775720517 Chr.17: 42852739-42852739 on Build GRCh38
rs11656660 Chr.17: 42852728-42852728 on Build GRCh38
rs199958009 Chr.17: 42852594-42852594 on Build GRCh38
rs753226579 Chr.17: 42852600-42852600 on Build GRCh38
rs11655622 Chr.17: 42852730-42852730 on Build GRCh38
rs767179590 Chr.17: 42852722-42852723 on Build GRCh38
rs202120479 Chr.17: 42852727-42852727 on Build GRCh38
rs535744858 Chr.17: 42852591-42852591 on Build GRCh38
rs115107156 Chr.17: 42852690-42852690 on Build GRCh38
rs33986943 Chr.17: 42852620-42852620 on Build GRCh38
rs201511383 Chr.17: 42852746-42852746 on Build GRCh38
rs112059383 Chr.17: 42852636-42852636 on Build GRCh38
rs201747345 Chr.17: 42852574-42852574 on Build GRCh38
rs762888597 Chr.17: 42852741-42852741 on Build GRCh38
rs145953403 Chr.17: 42852615-42852615 on Build GRCh38
rs374921494 Chr.17: 42852612-42852612 on Build GRCh38
rs769740181 Chr.17: 42852572-42852572 on Build GRCh38
rs771735567 Chr.17: 42852725-42852725 on Build GRCh38
rs763924559 Chr.17: 42852575-42852575 on Build GRCh38
rs748004654 Chr.17: 42852717-42852717 on Build GRCh38
rs780932168 Chr.17: 42852673-42852673 on Build GRCh38
rs780012474 Chr.17: 42852624-42852624 on Build GRCh38
rs530957915 Chr.17: 42852716-42852716 on Build GRCh38
rs201792517 Chr.17: 42852630-42852630 on Build GRCh38
rs777044564 Chr.17: 42852726-42852726 on Build GRCh38
rs544480745 Chr.17: 42852708-42852708 on Build GRCh38
rs745407800 Chr.17: 42852621-42852621 on Build GRCh38
rs562607668 Chr.17: 42852711-42852711 on Build GRCh38
rs754637660 Chr.17: 42852697-42852697 on Build GRCh38
rs753538898 Chr.17: 42852659-42852659 on Build GRCh38
rs141617446 Chr.17: 42852606-42852606 on Build GRCh38
rs201221953 Chr.17: 42852633-42852633 on Build GRCh38
rs368455364 Chr.17: 42852619-42852619 on Build GRCh38
rs765878171 Chr.17: 42852599-42852599 on Build GRCh38
rs367848102 Chr.17: 42852710-42852710 on Build GRCh38
rs759819332 Chr.17: 42852734-42852734 on Build GRCh38
rs555589985 Chr.17: 42852667-42852667 on Build GRCh38
rs141645485 Chr.17: 42852626-42852626 on Build GRCh38
rs753963358 Chr.17: 42852592-42852592 on Build GRCh38
rs2229596 Chr.17: 42852664-42852664 on Build GRCh38
rs757517219 Chr.17: 42852668-42852668 on Build GRCh38
rs751352031 Chr.17: 42852588-42852588 on Build GRCh38
rs372961818 Chr.17: 42852693-42852693 on Build GRCh38
rs753314218 Chr.17: 42852661-42852661 on Build GRCh38
rs35643019 Chr.17: 42852565-42852565 on Build GRCh38
rs764686944 Chr.17: 42852743-42852743 on Build GRCh38

View Full Product Details