Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 81109519-81109788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS3
Entrez Gene ID
79611
Gene Name
acyl-CoA synthetase short-chain family member 3
Gene Aliases
-
UniGene
Hs.259559
Gene Chromosome Location
Chr.12: 81078018-81257795 on Build GRCh38
Transcripts
NM_024560.2 XM_005269150.2 XM_005269151.4 XM_011538740.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4899031 c.405T>C p.S135S Chr.12: 81109653-81109653 on Build GRCh38
4848004 c.364G>C p.E122Q Chr.12: 81109612-81109612 on Build GRCh38
7285425 c.434C>T p.T145I Chr.12: 81109682-81109682 on Build GRCh38
431903 c.315T>C p.F105F Chr.12: 81109563-81109563 on Build GRCh38
1299900 c.333C>T p.N111N Chr.12: 81109581-81109581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780154222 Chr.12: 81109650-81109650 on Build GRCh38
rs553905670 Chr.12: 81109733-81109733 on Build GRCh38
rs368706037 Chr.12: 81109597-81109597 on Build GRCh38
rs1007154648 Chr.12: 81109671-81109671 on Build GRCh38
rs770447768 Chr.12: 81109596-81109596 on Build GRCh38
rs139156547 Chr.12: 81109549-81109549 on Build GRCh38
rs770630643 Chr.12: 81109687-81109687 on Build GRCh38
rs775039105 Chr.12: 81109717-81109717 on Build GRCh38
rs139584882 Chr.12: 81109631-81109631 on Build GRCh38
rs762948052 Chr.12: 81109751-81109751 on Build GRCh38
rs749183389 Chr.12: 81109591-81109591 on Build GRCh38
rs577406111 Chr.12: 81109698-81109698 on Build GRCh38
rs750597914 Chr.12: 81109639-81109639 on Build GRCh38
rs145208016 Chr.12: 81109642-81109642 on Build GRCh38
rs1016842077 Chr.12: 81109570-81109570 on Build GRCh38
rs867094815 Chr.12: 81109636-81109636 on Build GRCh38
rs777575475 Chr.12: 81109660-81109660 on Build GRCh38
rs746024523 Chr.12: 81109651-81109651 on Build GRCh38
rs779956713 Chr.12: 81109745-81109745 on Build GRCh38
rs760329091 Chr.12: 81109723-81109723 on Build GRCh38
rs780321014 Chr.12: 81109557-81109557 on Build GRCh38
rs368632922 Chr.12: 81109736-81109736 on Build GRCh38
rs767171201 Chr.12: 81109647-81109647 on Build GRCh38
rs760956945 Chr.12: 81109624-81109624 on Build GRCh38
rs770014944 Chr.12: 81109654-81109654 on Build GRCh38
rs747952146 Chr.12: 81109560-81109560 on Build GRCh38
rs759012460 Chr.12: 81109603-81109603 on Build GRCh38
rs1006445813 Chr.12: 81109755-81109755 on Build GRCh38
rs200270059 Chr.12: 81109691-81109691 on Build GRCh38
rs765902810 Chr.12: 81109742-81109742 on Build GRCh38
rs146304622 Chr.12: 81109604-81109604 on Build GRCh38
rs548439952 Chr.12: 81109581-81109581 on Build GRCh38
rs765809210 Chr.12: 81109635-81109635 on Build GRCh38
rs764481736 Chr.12: 81109626-81109626 on Build GRCh38
rs769574875 Chr.12: 81109575-81109575 on Build GRCh38
rs1026058341 Chr.12: 81109739-81109739 on Build GRCh38
rs371815204 Chr.12: 81109708-81109708 on Build GRCh38
rs749059659 Chr.12: 81109674-81109674 on Build GRCh38
rs374545940 Chr.12: 81109732-81109732 on Build GRCh38
rs371371273 Chr.12: 81109714-81109714 on Build GRCh38

View Full Product Details