Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57276507-57276768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB14
Entrez Gene ID
142686
Gene Name
ankyrin repeat and SOCS box containing 14
Gene Aliases
-
UniGene
Hs.665154
Gene Chromosome Location
Chr.3: 57268351-57293047 on Build GRCh38
Transcripts
NM_001142733.2 NM_130387.5 XM_017005736.1 XM_017005737.1 NR_135535.1

Gene Symbol
LOC105377102
Entrez Gene ID
105377102
Gene Name
uncharacterized LOC105377102
Gene Aliases
-
UniGene
Hs.745243
Gene Chromosome Location
Chr.3: 57269566-57278127 on Build GRCh38
Transcripts
NM_001142733.2 NM_130387.5 XM_017005736.1 XM_017005737.1 NR_135535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5527003 c.1742G>A p.G581E Chr.3: 57276572-57276572 on Build GRCh38
7400207 c.1701A>C p.K567N Chr.3: 57276613-57276613 on Build GRCh38
4621163 c.780C>G p.C260W Chr.3: 57276679-57276679 on Build GRCh38
6331112 c.1615C>T p.R539C Chr.3: 57276699-57276699 on Build GRCh38
1047624 c.754T>G p.L252V Chr.3: 57276705-57276705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142947683 Chr.3: 57276660-57276660 on Build GRCh38
rs377599276 Chr.3: 57276644-57276644 on Build GRCh38
rs146163375 Chr.3: 57276687-57276687 on Build GRCh38
rs1029579308 Chr.3: 57276585-57276585 on Build GRCh38
rs770387841 Chr.3: 57276592-57276592 on Build GRCh38
rs926651955 Chr.3: 57276656-57276656 on Build GRCh38
rs143429908 Chr.3: 57276630-57276630 on Build GRCh38
rs143068856 Chr.3: 57276623-57276623 on Build GRCh38
rs747745603 Chr.3: 57276622-57276622 on Build GRCh38
rs762743268 Chr.3: 57276698-57276698 on Build GRCh38
rs768894624 Chr.3: 57276616-57276616 on Build GRCh38
rs752546733 Chr.3: 57276651-57276651 on Build GRCh38
rs765047654 Chr.3: 57276557-57276557 on Build GRCh38
rs370017118 Chr.3: 57276609-57276609 on Build GRCh38
rs779936480 Chr.3: 57276717-57276717 on Build GRCh38
rs767635090 Chr.3: 57276686-57276686 on Build GRCh38
rs561259437 Chr.3: 57276562-57276562 on Build GRCh38
rs780889937 Chr.3: 57276729-57276729 on Build GRCh38
rs1005212361 Chr.3: 57276575-57276575 on Build GRCh38
rs772971599 Chr.3: 57276620-57276620 on Build GRCh38
rs898039892 Chr.3: 57276668-57276668 on Build GRCh38
rs574573222 Chr.3: 57276572-57276572 on Build GRCh38
rs759490763 Chr.3: 57276643-57276643 on Build GRCh38
rs147845490 Chr.3: 57276621-57276621 on Build GRCh38
rs746384971 Chr.3: 57276589-57276589 on Build GRCh38
rs373470439 Chr.3: 57276610-57276610 on Build GRCh38
rs756317617 Chr.3: 57276716-57276716 on Build GRCh38
rs140539832 Chr.3: 57276724-57276724 on Build GRCh38
rs750665339 Chr.3: 57276709-57276709 on Build GRCh38
rs563559307 Chr.3: 57276588-57276588 on Build GRCh38
rs267599914 Chr.3: 57276561-57276561 on Build GRCh38
rs200729395 Chr.3: 57276550-57276550 on Build GRCh38
rs543255867 Chr.3: 57276580-57276580 on Build GRCh38
rs779355965 Chr.3: 57276720-57276720 on Build GRCh38
rs141916461 Chr.3: 57276722-57276722 on Build GRCh38
rs752487875 Chr.3: 57276559-57276559 on Build GRCh38
rs769290213 Chr.3: 57276688-57276688 on Build GRCh38
rs868348729 Chr.3: 57276726-57276727 on Build GRCh38
rs763957241 Chr.3: 57276699-57276699 on Build GRCh38
rs774511412 Chr.3: 57276679-57276679 on Build GRCh38
rs745897762 Chr.3: 57276670-57276670 on Build GRCh38
rs776078499 Chr.3: 57276636-57276636 on Build GRCh38
rs745507743 Chr.3: 57276738-57276738 on Build GRCh38
rs751276157 Chr.3: 57276703-57276703 on Build GRCh38
rs781456578 Chr.3: 57276664-57276664 on Build GRCh38
rs150457384 Chr.3: 57276659-57276659 on Build GRCh38
rs555966043 Chr.3: 57276663-57276663 on Build GRCh38
rs868086701 Chr.3: 57276727-57276728 on Build GRCh38
rs375766030 Chr.3: 57276705-57276705 on Build GRCh38
rs762153247 Chr.3: 57276704-57276704 on Build GRCh38
rs370930102 Chr.3: 57276694-57276694 on Build GRCh38
rs563892101 Chr.3: 57276611-57276611 on Build GRCh38
rs199925423 Chr.3: 57276671-57276671 on Build GRCh38
rs769076047 Chr.3: 57276677-57276677 on Build GRCh38
rs144656144 Chr.3: 57276719-57276719 on Build GRCh38

View Full Product Details