Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87409189-87409462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB4
Entrez Gene ID
5244
Gene Name
ATP binding cassette subfamily B member 4
Gene Aliases
ABC21, GBD1, ICP3, MDR2, MDR2/3, MDR3, PFIC-3, PGY3
UniGene
Hs.654403
Gene Chromosome Location
Chr.7: 87399461-87475864 on Build GRCh38
Transcripts
NM_000443.3 NM_018849.2 NM_018850.2 XM_011516308.2 XM_011516309.2 XM_011516310.2 XM_011516311.2 XM_011516312.2 XM_011516313.2 XM_011516315.2 XM_017012323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5428979 c.3006G>T p.L1002L Chr.7: 87409311-87409311 on Build GRCh38
3703301 c.2925-8C>A p.? Chr.7: 87409400-87409400 on Build GRCh38
6589995 c.2957C>T p.A986V Chr.7: 87409360-87409360 on Build GRCh38
3883034 c.3074T>C p.L1025P Chr.7: 87409243-87409243 on Build GRCh38
336930 c.2965C>A p.H989N Chr.7: 87409352-87409352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745753830 Chr.7: 87409418-87409418 on Build GRCh38
rs140948592 Chr.7: 87409331-87409331 on Build GRCh38
rs752863097 Chr.7: 87409396-87409396 on Build GRCh38
rs1005279302 Chr.7: 87409406-87409406 on Build GRCh38
rs867322580 Chr.7: 87409239-87409239 on Build GRCh38
rs928701624 Chr.7: 87409276-87409276 on Build GRCh38
rs765729147 Chr.7: 87409228-87409228 on Build GRCh38
rs370644411 Chr.7: 87409379-87409379 on Build GRCh38
rs531111327 Chr.7: 87409304-87409304 on Build GRCh38
rs45574932 Chr.7: 87409365-87409365 on Build GRCh38
rs756238268 Chr.7: 87409400-87409400 on Build GRCh38
rs779028005 Chr.7: 87409409-87409409 on Build GRCh38
rs776190374 Chr.7: 87409332-87409332 on Build GRCh38
rs758770046 Chr.7: 87409238-87409238 on Build GRCh38
rs773701919 Chr.7: 87409256-87409256 on Build GRCh38
rs750829010 Chr.7: 87409235-87409235 on Build GRCh38
rs538954855 Chr.7: 87409260-87409260 on Build GRCh38
rs766698345 Chr.7: 87409373-87409373 on Build GRCh38
rs755016053 Chr.7: 87409267-87409267 on Build GRCh38
rs748220777 Chr.7: 87409286-87409286 on Build GRCh38
rs970022822 Chr.7: 87409244-87409244 on Build GRCh38
rs948902384 Chr.7: 87409364-87409364 on Build GRCh38
rs771055973 Chr.7: 87409290-87409290 on Build GRCh38
rs149043617 Chr.7: 87409271-87409271 on Build GRCh38
rs372458542 Chr.7: 87409426-87409426 on Build GRCh38
rs549345313 Chr.7: 87409323-87409323 on Build GRCh38
rs374711909 Chr.7: 87409381-87409381 on Build GRCh38
rs1023568209 Chr.7: 87409236-87409236 on Build GRCh38
rs765784054 Chr.7: 87409348-87409348 on Build GRCh38
rs111763305 Chr.7: 87409353-87409353 on Build GRCh38
rs1051861187 Chr.7: 87409385-87409385 on Build GRCh38
rs761283092 Chr.7: 87409333-87409333 on Build GRCh38
rs367934441 Chr.7: 87409388-87409388 on Build GRCh38
rs771863019 Chr.7: 87409307-87409307 on Build GRCh38
rs56187107 Chr.7: 87409370-87409370 on Build GRCh38
rs1057495486 Chr.7: 87409317-87409317 on Build GRCh38
rs760321007 Chr.7: 87409327-87409327 on Build GRCh38
rs780160089 Chr.7: 87409248-87409248 on Build GRCh38
rs2230029 Chr.7: 87409280-87409280 on Build GRCh38

View Full Product Details