Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62805173-62805414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG4C
Entrez Gene ID
84938
Gene Name
autophagy related 4C cysteine peptidase
Gene Aliases
APG4-C, APG4C, AUTL1, AUTL3
UniGene
Hs.7353
Gene Chromosome Location
Chr.1: 62784106-62865270 on Build GRCh38
Transcripts
NM_032852.3 NM_178221.2 XM_005271288.2 XM_011542313.2 XM_017002600.1 XM_017002601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
911429 c.137A>C p.K46T Chr.1: 62805232-62805232 on Build GRCh38
681528 c.96G>C p.K32N Chr.1: 62805191-62805191 on Build GRCh38
5716339 c.134G>A p.G45E Chr.1: 62805229-62805229 on Build GRCh38
5716338 c.134G>A p.G45E Chr.1: 62805229-62805229 on Build GRCh38
681529 c.96G>C p.K32N Chr.1: 62805191-62805191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143982322 Chr.1: 62805225-62805225 on Build GRCh38
rs773005472 Chr.1: 62805287-62805287 on Build GRCh38
rs965593150 Chr.1: 62805281-62805281 on Build GRCh38
rs770154953 Chr.1: 62805272-62805272 on Build GRCh38
rs1047505101 Chr.1: 62805211-62805213 on Build GRCh38
rs747599823 Chr.1: 62805226-62805226 on Build GRCh38
rs774222745 Chr.1: 62805276-62805276 on Build GRCh38
rs367704211 Chr.1: 62805222-62805222 on Build GRCh38
rs565870538 Chr.1: 62805320-62805320 on Build GRCh38
rs967173986 Chr.1: 62805357-62805357 on Build GRCh38
rs201685263 Chr.1: 62805251-62805251 on Build GRCh38
rs761545706 Chr.1: 62805284-62805284 on Build GRCh38
rs746075510 Chr.1: 62805261-62805261 on Build GRCh38
rs779188621 Chr.1: 62805214-62805214 on Build GRCh38
rs760709859 Chr.1: 62805292-62805292 on Build GRCh38
rs10789128 Chr.1: 62805339-62805339 on Build GRCh38
rs908941645 Chr.1: 62805350-62805350 on Build GRCh38
rs772000085 Chr.1: 62805286-62805286 on Build GRCh38
rs1000886632 Chr.1: 62805367-62805367 on Build GRCh38
rs1002829640 Chr.1: 62805256-62805256 on Build GRCh38
rs898753629 Chr.1: 62805250-62805250 on Build GRCh38
rs770883310 Chr.1: 62805241-62805241 on Build GRCh38
rs984496921 Chr.1: 62805346-62805346 on Build GRCh38
rs1007000689 Chr.1: 62805215-62805215 on Build GRCh38
rs1032726537 Chr.1: 62805369-62805369 on Build GRCh38
rs547561580 Chr.1: 62805300-62805300 on Build GRCh38
rs529086362 Chr.1: 62805227-62805227 on Build GRCh38
rs776129641 Chr.1: 62805304-62805304 on Build GRCh38

View Full Product Details