Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96551729-96551960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID5A
Entrez Gene ID
10865
Gene Name
AT-rich interaction domain 5A
Gene Aliases
MRF-1, MRF1, RP11-363D14
UniGene
Hs.920
Gene Chromosome Location
Chr.2: 96536719-96552638 on Build GRCh38
Transcripts
NM_001319085.1 NM_001319087.1 NM_001319092.1 NM_001319093.1 NM_001319094.1 NM_001319096.1 NM_212481.2 XM_011510499.2 XM_017003183.1 XM_017003184.1 XM_017003185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5619827 c.1234A>T p.K412* Chr.2: 96551762-96551762 on Build GRCh38
1023921 c.1269G>T p.K423N Chr.2: 96551797-96551797 on Build GRCh38
5059130 c.1378C>T p.R460W Chr.2: 96551906-96551906 on Build GRCh38
7191994 c.1406C>T p.A469V Chr.2: 96551934-96551934 on Build GRCh38
2825610 c.1335C>T p.R445R Chr.2: 96551863-96551863 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143598492 Chr.2: 96551775-96551775 on Build GRCh38
rs747001402 Chr.2: 96551771-96551771 on Build GRCh38
rs779502556 Chr.2: 96551887-96551887 on Build GRCh38
rs759670669 Chr.2: 96551817-96551817 on Build GRCh38
rs758229260 Chr.2: 96551762-96551762 on Build GRCh38
rs150726674 Chr.2: 96551870-96551870 on Build GRCh38
rs752970311 Chr.2: 96551819-96551819 on Build GRCh38
rs752421977 Chr.2: 96551748-96551748 on Build GRCh38
rs751362300 Chr.2: 96551858-96551858 on Build GRCh38
rs747413578 Chr.2: 96551907-96551907 on Build GRCh38
rs777135656 Chr.2: 96551912-96551912 on Build GRCh38
rs763975514 Chr.2: 96551828-96551828 on Build GRCh38
rs780831070 Chr.2: 96551776-96551776 on Build GRCh38
rs940372901 Chr.2: 96551824-96551824 on Build GRCh38
rs761191111 Chr.2: 96551809-96551809 on Build GRCh38
rs145972469 Chr.2: 96551789-96551789 on Build GRCh38
rs536788603 Chr.2: 96551934-96551934 on Build GRCh38
rs772273506 Chr.2: 96551902-96551902 on Build GRCh38
rs139848963 Chr.2: 96551862-96551862 on Build GRCh38
rs773876068 Chr.2: 96551861-96551861 on Build GRCh38
rs915277728 Chr.2: 96551838-96551838 on Build GRCh38
rs200341137 Chr.2: 96551885-96551885 on Build GRCh38
rs777507265 Chr.2: 96551766-96551766 on Build GRCh38
rs769562008 Chr.2: 96551790-96551790 on Build GRCh38
rs771414555 Chr.2: 96551911-96551911 on Build GRCh38
rs921845830 Chr.2: 96551811-96551811 on Build GRCh38
rs762023972 Chr.2: 96551798-96551798 on Build GRCh38
rs374390790 Chr.2: 96551906-96551906 on Build GRCh38
rs376260443 Chr.2: 96551823-96551823 on Build GRCh38
rs773550020 Chr.2: 96551806-96551806 on Build GRCh38
rs920373074 Chr.2: 96551927-96551927 on Build GRCh38
rs552761363 Chr.2: 96551778-96551778 on Build GRCh38
rs978552629 Chr.2: 96551772-96551772 on Build GRCh38
rs765385238 Chr.2: 96551818-96551818 on Build GRCh38
rs765390396 Chr.2: 96551917-96551917 on Build GRCh38
rs753882600 Chr.2: 96551814-96551814 on Build GRCh38
rs758085829 Chr.2: 96551813-96551813 on Build GRCh38
rs764267450 Chr.2: 96551935-96551935 on Build GRCh38
rs748999830 Chr.2: 96551888-96551888 on Build GRCh38
rs775267535 Chr.2: 96551797-96551797 on Build GRCh38
rs775529696 Chr.2: 96551922-96551922 on Build GRCh38

View Full Product Details